EMAIL THIS PAGE TO A FRIEND
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment
Multi-residue analysis of pesticides in traditional Chinese medicines using gas chromatography-negative chemical ionisation tandem mass spectrometry.
PMID 26125677
Abstract
In this study, a residue analysis method for the simultaneous determination of 107 pesticides in traditional Chinese medicines (TCMs), Angelica sinensis, A. dahurica, Leonurus heterophyllus Sweet, Pogostemon cablin and Lonicera japonica Thunb., was developed using gas chromatography coupled with tandem mass spectrometry in negative chemical ionisation mode (GC-NCI-MS/MS). NCI has advantages of high sensitivity and selectivity to chemicals with electron-withdrawing groups, and yields low background interference. For sample preparation, QuEChERS (quick, easy, cheap, effective, rugged and safe) was applied. Due to the unique characteristics of TCMs, the clean-up step was optimised by adjusting amounts of primary secondary amine, C18, graphitised carbon black and silica sorbents. Validation was mainly performed by determining analyte recoveries at four different spiking concentrations of 10, 50, 100 and 200xa0ngxa0g(-1), with seven replicates at each concentration. Method trueness, precision, linearity of calibration curves, lowest calibrated levels (LCLs) and matrix effects were determined to demonstrate method and instrument performance. Among the 107 pesticides tested, approximately 80% gave recoveries from 80% to 110% and