EMAIL THIS PAGE TO A FRIEND
ASN neuro
Combined HDAC1 and HDAC2 Depletion Promotes Retinal Ganglion Cell Survival After Injury Through Reduction of p53 Target Gene Expression.
PMID 26129908
Abstract
Histones deacetylases (HDACs), besides their function as epigenetic regulators, deacetylate and critically regulate the activity of nonhistone targets. In particular, HDACs control partially the proapoptotic activity of p53 by balancing its acetylation state. HDAC inhibitors have revealed neuroprotective properties in different models, but the exact mechanisms of action remain poorly understood. We have generated a conditional knockout mouse model targeting retinal ganglion cells (RGCs) to investigate specifically the functional role of HDAC1 and HDAC2 in an acute model of optic nerve injury. Our results demonstrate that combined HDAC1 and HDAC2 ablation promotes survival of axotomized RGCs. Based on global gene expression analyses, we identified the p53-PUMA apoptosis-inducing axis to be strongly activated in axotomized mouse RGCs. Specific HDAC1/2 ablation inhibited this apoptotic pathway by impairing the crucial acetylation status of p53 and reducing PUMA expression, thereby contributing to the ensuing enhanced neuroprotection due to HDAC1/2 depletion. HDAC1/2 inhibition and the affected downstream signaling components emerge as specific targets for developing therapeutic strategies in neuroprotection.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2501536
Anti-BBC3 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
J104
Anti-c-Jun antibody produced in rabbit, affinity isolated antibody, lyophilized powder
HPA035775
Anti-D4S234E antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2105613
Anti-D4S234E, (N-terminal) antibody produced in rabbit, affinity isolated antibody
GW22766 Anti-FAS (ab1) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
GW22767 Anti-FAS (ab2) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB2501317
Anti-FAS antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA027444
Anti-FAS antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2105218
Anti-FAS antibody produced in rabbit, affinity isolated antibody
SAB4501532
Anti-FAS, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4501533
Anti-FAS, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB2501930
Anti-HDAC1 (C-terminal) antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2502051
Anti-Hdac1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2701686
Anti-hdac1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701822
Anti-hdac1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
AV38530
Anti-HDAC1 antibody produced in rabbit, IgG fraction of antiserum
SAB2101020
Anti-HDAC1 antibody produced in rabbit, affinity isolated antibody
HPA029693
Anti-HDAC1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4501382
Anti-HDAC1, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4501383
Anti-HDAC1, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4300412
Anti-HDAC2 (Ab-394) antibody produced in rabbit, affinity isolated antibody
HPA011727
Anti-HDAC2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV32618
Anti-HDAC2 antibody produced in rabbit, affinity isolated antibody
SAB2104633
Anti-HDAC2 antibody produced in rabbit, affinity isolated antibody
SAB4501384
Anti-HDAC2, C-Terminal antibody produced in rabbit, affinity isolated antibody
H3284
Anti-Histone Deacetylase 1 (HDAC1) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
H6287 Anti-Histone Deacetylase 1 (HDAC1) antibody, Mouse monoclonal, clone HDAC1-21, purified from hybridoma cell culture
H3159
Anti-Histone Deacetylase 2 (HDAC2) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
H2663
Anti-Histone Deacetylase 2 (HDAC2) antibody, Mouse monoclonal, clone HDAC2-62, purified from hybridoma cell culture
SAB4200176
Anti-JNK antibody, Mouse monoclonal, clone 1C2, purified from hybridoma cell culture