EMAIL THIS PAGE TO A FRIEND
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine
Carbonic anhydrase IX overexpression regulates the migration and progression in oral squamous cell carcinoma.
PMID 26130414
Abstract
Carbonic anhydrase IX (CAIX) is reportedly overexpressed in several types of carcinomas and is generally considered a marker of malignancy. The current study investigated the association between membrane expression of CAIX and the clinicopathological characteristics in oral squamous cell carcinoma (OSCC) patients. The study used immunohistochemistry to examine CAIX expression in 271 OSCC specimens by tissue microarray (TMA) and assessed the effect of CAIX overexpression and knockdown on migration of oral cancer cells in vitro. We found that CAIX expression was associated with more advanced clinical stages (p = 0.030) and positive lymph node metastasis (p = 0.026). Importantly, CAIX expression was correlated with a poorer patient prognosis in a univariate survival analysis (p = 0.025). Moreover, CAIX suppression by small interfering RNA (siRNA) significantly reduced cellular migration in OECM-1 oral cancer cell. In conclusion, our study showed that the expression of CAIX in OSCC samples can predict the progression of OSCC and survival of OSCC patients in Taiwan.