EMAIL THIS PAGE TO A FRIEND
Environmental science & technology
Effects of Simulated Rare Earth Recycling Wastewaters on Biological Nitrification.
PMID 26132866
Abstract
Increasing rare earth element (REE) supplies by recycling and expanded ore processing will result in generation of new wastewaters. In some cases, disposal to a sewage treatment plant may be favored, but plant performance must be maintained. To assess the potential effects of such wastewaters on biological treatment, model nitrifying organisms Nitrosomonas europaea and Nitrobacter winogradskyi were exposed to simulated wastewaters containing varying levels of yttrium or europium (10, 50, and 100 ppm), and the extractant tributyl phosphate (TBP, at 0.1 g/L). Y and Eu additions at 50 and 100 ppm inhibited N. europaea, even when virtually all of the REE was insoluble. Provision of TBP with Eu increased N. europaea inhibition, although TBP alone did not substantially alter activity. For N. winogradskyi cultures, Eu or Y additions at all tested levels induced significant inhibition, and nitrification shut down completely with TBP addition. REE solubility was calculated using the previously developed MSE (Mixed-Solvent Electrolyte) thermodynamic model. The model calculations reveal a strong pH dependence of solubility, typically controlled by the precipitation of REE hydroxides but also likely affected by the formation of unknown phosphate phases, which determined aqueous concentrations experienced by the microorganisms.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
429732
Europium(III) chloride, anhydrous, powder, 99.99% trace metals basis
Cl3Eu
238066
Europium(III) chloride, powder, ≥99.9% trace metals basis
Cl3Eu
158615
Tributyl phosphate, 97%
C12H27O4P
240494
Tributyl phosphate, ≥99%
C12H27O4P
262994
Yttrium, chips, 99.9% trace rare earth metals basis
Y
451347
Yttrium, dendritic pieces, 99.9% trace metals basis
Y
693642
Yttrium, foil, thickness 1 mm, 99.8% trace rare earth metals basis
Y
261319
Yttrium, ingot, 99.9% trace rare earth metals basis
Y
261327
Yttrium, powder, −40 mesh, 99.5% trace rare earth metals basis
Y
GF09119279
Yttrium, foil, 25x25mm, thickness 0.15mm, as rolled, 99%
Y
GF77674719
Yttrium, wire reel, 0.2m, diameter 0.5mm, 99.9%
Y
GF11301398
Yttrium, rod, 50mm, diameter 6.35mm, cast, 99%
Y
GF86394156
Yttrium, wire reel, 0.2m, diameter 1.0mm, 99.9%
Y
GF91377478
Yttrium, foil, 25x25mm, thickness 0.125mm, as rolled, 99%
Y
GF87149481
Yttrium, foil, 50x50mm, thickness 0.125mm, as rolled, 99%
Y
GF03037148
Yttrium, foil, 50x50mm, thickness 0.15mm, as rolled, 99%
Y
GF55118257
Yttrium, foil, 25mm disks, thickness 0.50mm, as rolled, 99%
Y
GF33100298
Yttrium, foil, not light tested, 100x100mm, thickness 0.025mm, as rolled, 99%
Y
GF75315719
Yttrium, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99%
Y
GF61507740
Yttrium, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99%
Y
GF40879008
Yttrium, rod, 100mm, diameter 12.5mm, cast, 99%
Y
GF12674617
Yttrium, rod, 100mm, diameter 6.35mm, cast, 99%
Y
GF84327037
Yttrium, rod, 50mm, diameter 12.5mm, cast, 99%
Y
GF68048860
Yttrium, rod, 50mm, diameter 2.0mm, cast, 99%
Y
GF68472692
Yttrium, wire reel, 0.05m, diameter 0.5mm, 99.9%
Y
GF35512725
Yttrium, wire reel, 0.05m, diameter 1.0mm, 99.9%
Y
GF95617020
Yttrium, wire reel, 0.05m, diameter 1.0mm, square section, 99.9%
Y
GF02763963
Yttrium, wire reel, 0.1m, diameter 0.5mm, 99.9%
Y
GF39091158
Yttrium, wire reel, 0.1m, diameter 1.0mm, 99.9%
Y
GF79949255
Yttrium, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99%
Y