EMAIL THIS PAGE TO A FRIEND
Oncology reports
Trastuzumab and docetaxel in a preclinical organotypic breast cancer model using tissue slices from mammary fat pad: Translational relevance.
PMID 26133490
Abstract
With the ever-increasing number of drugs approved to treat cancers, selection of the optimal treatment regimen for an individual patient is challenging. Breast cancer complexity requires novel predictive methods and tools. In the present study, we set up experimental conditions to obtain an 'exxa0vivo' organotypic culture from xenotransplanted mice aiming at recapitulating the human clinical condition. The effect of trastuzumab (large biological molecule) and docetaxel (small chemical entity) was subsequently investigated on this organotypic model and compared with inxa0vivo and inxa0vitro activity on tumor cells. Tissue slices of 200xa0µm were obtained from mammary fat pad of SCID mice xenotransplanted with human MCF-7 breast cancer cells. Viability and proliferation were evaluated by 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyl-tetrazolium bromide (MTT) colorimetric assay and Ki-67 immunohistochemistry,and apoptosis by cleaved caspase-3 immunohistochemistry. Inxa0vivo antitumor activity of trastuzumab and docetaxel was determined by caliper measurement of tumor volume and Ki-67 expression on explanted masses by immunohistochemistry. A Teflon support and normoxia were necessary experimental conditions to obtain high viability of excised breast cancer infiltrated mammary fat pad slices upon 48xa0h cultivation, as shown by MTT proliferation assay, and Ki-67 expression. Breast cancer tissue slices treated for 48xa0h with trastuzumab or docetaxel showed a significant dose‑dependent reduction of viability by MTT assay. Consistently, both drugs down-modulated Ki-67 and increased cleaved caspase-3. Tumor masses collected from docetaxel- or trastuzumab‑treated mice showed a similar reduction of proliferation markers. By contrast, MCF-7 cell cultures were significantly inhibited by docetaxel but not by trastuzumab. Tumor tissue slices represent a more predictive experimental cancer model compared to cell cultures for both small and large molecule antitumor efficacy. This observation supports the relevance of microenvironment in the overall tumor biology and response to therapeutics.