EMAIL THIS PAGE TO A FRIEND
Oncology reports
Sulforaphane inhibits invasion by phosphorylating ERK1/2 to regulate E-cadherin and CD44v6 in human prostate cancer DU145 cells.
PMID 26134113
Abstract
Advanced prostate cancer has highly invasive potential, which may lead to metastasis associated with poor prognosis. Sulforaphane (SFN), abundant in cruciferous vegetables, exhibited effective resistance to carcinogenesis in a variety of tumors. The aim of the present study was to investigate whether SFN inhibited invasion in human prostate cancer cells via sustained activation of ERK1/2 and downstream signaling by an invasion assay, gelatin zymography and western blot analysis. The results showed that SFN inhibited invasion and we characterized the underlying mechanisms in human DU145 prostate cancer cells. SFN (15xa0µM) changed cell morphology leading to short‑cell pseudopodia which may suppress tumor migration and invasion. The Transwell assay showed that SFN phosphorylated ERK1/2 in a dose- and time-dependent manner and significantly inhibited cell invasion, while the effect was reduced by the ERK1/2 blocker PD98059 (25xa0µM). Furthermore, these effects contributed to the upregulation of E-cadherin and the downregulation of CD44v6 and were eradicated by PD98059. Western blot analysis and gelatin zymography showed that SFN decreased the expression and activity of MMP-2. Thus, SFN inhibited invasion by activating ERK1/2 to upregulate E-cadherin and downregulate CD44v6, thereby reducing MMP-2 expression and activity. E-cadherin is an invasion inhibitor, while CD44v6 and MMP-2 are invasion promoters. Therefore, SFN is a prospective therapeutic agent that may be used to prevent invasion in prostate cancer.