EMAIL THIS PAGE TO A FRIEND
Clinical and vaccine immunology : CVI
Human Neutrophil Lipocalin as a Superior Diagnostic Means To Distinguish between Acute Bacterial and Viral Infections.
PMID 26135974
Abstract
The distinction between causes of acute infections is a major clinical challenge. Current biomarkers, however, are not sufficiently accurate. Human neutrophil lipocalin (HNL) concentrations in serum or whole blood activated by formyl-methionine-leucine-phenylalanine (fMLP) were shown to distinguish acute infections of bacterial or viral cause with high accuracy. The aim was therefore to compare the clinical performance of HNL with currently used biomarkers. Seven hundred twenty-five subjects (144 healthy controls and 581 patients with signs and symptoms of acute infections) were included in the study. C-reactive protein (CRP), the expression of CD64 on neutrophils, procalcitonin (PCT), and blood neutrophil counts were measured by established techniques, and HNL concentrations were measured in whole-blood samples after activation with fMLP. All tested biomarkers were elevated in bacterial as opposed to viral infections (P < 0.001). CRP, PCT, and CD64 expression in neutrophils was elevated in viral infections compared to healthy controls (P < 0.001). In the distinction between healthy controls and patients with bacterial infections, the areas under the receiver operating characteristic (ROC) curves were >0.85 for all biomarkers, whereas for the distinction between bacterial and viral infections, only HNL concentration in fMLP-activated whole blood showed an area under the ROC curve (AUROC) of >0.90 and superior clinical performance. The clinical performance of HNL in fMLP-activated whole blood was superior to current biomarkers and similar to previous results of HNL in serum. The procedure can be adopted for point-of-care testing with response times of <15 min.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W372609
DL-Phenylalanine, 99%, FCC
C9H11NO2