EMAIL THIS PAGE TO A FRIEND
Journal of cell science
Transport of the cholera toxin B-subunit from recycling endosomes to the Golgi requires clathrin and AP-1.
PMID 26136365
Abstract
The retrograde pathway is defined by the transport of proteins and lipids from the plasma membrane through endosomes to the Golgi complex, and is essential for a variety of cellular activities. Recycling endosomes are important sorting stations for some retrograde cargo. SMAP2, a GTPase-activating protein (GAP) for Arf1 with a putative clathrin-binding domain, has previously been shown to participate in the retrograde transport of the cholera toxin B-subunit (CTxB) from recycling endosomes. Here, we found that clathrin, a vesicle coat protein, and clathrin adaptor protein complex 1 (AP-1) were present at recycling endosomes and were needed for the retrograde transport of CTxB from recycling endosomes to the Golgi, but not from the plasma membrane to recycling endosomes. SMAP2 immunoprecipitated clathrin and AP-1 through a putative clathrin-binding domain and a CALM-binding domain, and SMAP2 mutants that did not interact with clathrin or AP-1 could not localize to recycling endosomes. Moreover, knockdown of Arf1 suppressed the retrograde transport of CTxB from recycling endosomes to the Golgi. These findings suggest that retrograde transport is mediated by clathrin-coated vesicles from recycling endosomes and that the role of the coat proteins is in the recruitment of Arf GAP to transport vesicles.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA001841
Anti-BRD8 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2100223
Anti-BRD8 antibody produced in rabbit, affinity isolated antibody
HPA043280
Anti-CLINT1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1305405
ANTI-ENTH (CENTER) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
G7295
Anti-GM130 (C-terminal) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1405886
Anti-GOLGA2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA021178
Anti-GOLGA2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
HPA021230
Anti-GOLGA2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
HPA021799
Anti-GOLGA2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab3
SAB3701093 Anti-Mouse IgG (H+L) antibody produced in sheep, affinity isolated antibody, buffered aqueous solution
SAB1307011
ANTI-SMAP2 (CENTER) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA021466
Anti-SMAP2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
HPA024424
Anti-SMAP2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
SAB1304463
ANTI-SMOC2 (C-TERM) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB2700686
Anti-SMOC2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA001358
Anti-STX5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2701187
Anti-STX5 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA001527
Anti-TF antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA005692
Anti-TF antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
T9826
Anti-TGN38 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
T7576
Anti-TGN46 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
HPA012609
Anti-TGOLN2 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA012723
Anti-TGOLN2 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2103825
Anti-TGOLN2, (N-terminal) antibody produced in rabbit, affinity isolated antibody
GW20009F
Anti-Transferrin antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
T2027 Anti-Transferrin antibody produced in goat, whole antiserum
SAB2700982
Anti-VPS35 (C-terminal) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701269
Anti-VPS35 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA040802
Anti-VPS35 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
EHU010871 MISSION® esiRNA, esiRNA targeting human CLINT1 (esiRNA1)