EMAIL THIS PAGE TO A FRIEND
Journal of virology
The Intracellular Domain of the Coxsackievirus and Adenovirus Receptor Differentially Influences Adenovirus Entry.
PMID 26136571
Abstract
The coxsackievirus and adenovirus receptor (CAR) is a cell adhesion molecule used as a docking molecule by some adenoviruses (AdVs) and group B coxsackieviruses. We previously proposed that the preferential transduction of neurons by canine adenovirus type 2 (CAV-2) is due to CAR-mediated internalization. Our proposed pathway of CAV-2 entry is in contrast to that of human AdV type 5 (HAdV-C5) in nonneuronal cells, where internalization is mediated by auxiliary receptors such as integrins. We therefore asked if in fibroblast-like cells the intracellular domain (ICD) of CAR plays a role in the internalization of the CAV-2 fiber knob (FK(CAV)), CAV-2, or HAdV-C5 when the capsid cannot engage integrins. Here, we show that in fibroblast-like cells, the CAR ICD is needed for FK(CAV) entry and efficient CAV-2 transduction but dispensable for HAdV-C5 and an HAdV-C5 capsid lacking the RGD sequence (an integrin-interacting motif) in the penton. Moreover, the deletion of the CAR ICD further impacts CAV-2 intracellular trafficking, highlighting the crucial role of CAR in CAV-2 intracellular dynamics. These data demonstrate that the CAR ICD contains sequences important for the recruitment of the endocytic machinery that differentially influences AdV cell entry. Understanding how viruses interact with the host cell surface and reach the intracellular space is of crucial importance for applied and fundamental virology. Here, we compare the role of a cell adhesion molecule (CAR) in the internalization of adenoviruses that naturally infect humans and Canidae. We show that the intracellular domain of CAR differentially regulates AdV entry and trafficking. Our study highlights the mechanistic differences that a receptor can have for two viruses from the same family.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB3500285
Anti-LAMP-1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
L1418
Anti-LAMP1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA014750
Anti-LAMP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1411780
ANTI-RCV1 antibody produced in mouse, clone 5A6, purified immunoglobulin, buffered aqueous solution
SAB1411781
ANTI-RCV1 antibody produced in mouse, clone 2H4, purified immunoglobulin, buffered aqueous solution
SAB1411794
ANTI-RCV1 antibody produced in mouse, clone 4H2, purified immunoglobulin, buffered aqueous solution
SAB1411795
ANTI-RCV1 antibody produced in mouse, clone 4C10, purified immunoglobulin, buffered aqueous solution
SAB1411804
ANTI-RCV1 antibody produced in mouse, clone 4D9, purified immunoglobulin, buffered aqueous solution
SAB1411805
ANTI-RCV1 antibody produced in mouse, clone 2B3, purified immunoglobulin, buffered aqueous solution
SAB1411862
ANTI-RCV1 antibody produced in mouse, clone 1D3, purified immunoglobulin, buffered aqueous solution
SAB1411863
ANTI-RCV1 antibody produced in mouse, clone 4E2, purified immunoglobulin, buffered aqueous solution
SAB1411864
ANTI-RCV1 antibody produced in mouse, clone 4A8, purified immunoglobulin, buffered aqueous solution
SAB1411865
ANTI-RCV1 antibody produced in mouse, clone 2B2, purified immunoglobulin, buffered aqueous solution
SAB1411878
ANTI-RCV1 antibody produced in mouse, clone 1B9, purified immunoglobulin, buffered aqueous solution
SAB1411879
ANTI-RCV1 antibody produced in mouse, clone 1B7, purified immunoglobulin, buffered aqueous solution
AV38196
Anti-RCV1 antibody produced in rabbit, affinity isolated antibody
SAB1406380
Anti-RCVRN antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2101971
Anti-RCVRN antibody produced in rabbit, affinity isolated antibody
SAB4200309
Anti-Recoverin (C-terminal) antibody produced in rabbit, ~1.5 mg/mL, affinity isolated antibody
SAB4502604
Anti-Recoverin antibody produced in rabbit, affinity isolated antibody
WH0005957M1
Monoclonal Anti-RCV1 antibody produced in mouse, clone 4C6, purified immunoglobulin, buffered aqueous solution
SAB1404306
Monoclonal Anti-RCV1, (C-terminal) antibody produced in mouse, clone 3G10, purified immunoglobulin, buffered aqueous solution