EMAIL THIS PAGE TO A FRIEND
Mycopathologia
Agar Diffusion Procedures for Susceptibility Testing of Malassezia pachydermatis: Evaluation of Mueller-Hinton Agar Plus 2 % Glucose and 0.5 µg/ml Methylene Blue as the Test Medium.
PMID 26138434
Abstract
Aim of this study was to verify whether Mueller-Hinton agar supplemented with 2 % glucose and methylene blue (MH-GM), which is used for disk diffusion susceptibility testing of Candida species by the Clinical and Laboratory Standards Institute, is suitable for testing Malassezia pachydermatis. A variant of the disk diffusion procedure utilizing a 9-mm tablet was used to test 31 isolates against clotrimazole and miconazole using MH-GM as test medium. The MH-GM agar optimally supported the growth of all M. pachydermatis isolates, provided that the yeast inoculum was prepared with a lipid source (Tween 40 and 80). Zone edges were frequently definite and clear, facilitating the measurement of zone size and minimizing subjectivity. The inhibition zones correlated with MIC values obtained in a broth dilution assay. The agar diffusion method with MH-GM as the test medium appears as a suitable procedure for testing the susceptibility of M. pachydermatis to CTZ and MCZ in clinical laboratories. This test format may allow processing a large number of isolates in epidemiological studies. This may in turn facilitate clarifying to what extent the problem "drug resistance" accounts for cases of treatment failure in dogs with Malassezia otitis and dermatitis.