EMAIL THIS PAGE TO A FRIEND
Cytotherapy
Useful properties of undifferentiated mesenchymal stromal cells and adipose tissue as the source in liver-regenerative therapy studied in an animal model of severe acute fulminant hepatitis.
PMID 26139545
Abstract
End-stage liver diseases frequently require liver transplantation. Cell therapy could be an alternative. This study aimed to analyze whether undifferentiated mesenchymal stromal cells (U-MSCs) or MSC-derived hepatocyte-like cells (DHLCs) from adipose tissue (AT), umbilical cord blood (UCB) and bone marrow (BM) would better restore damaged liver. AT was obtained from lipo-aspiration, UCB from an Umbilical Cord Blood Bank and BM from a BM Transplantation Unit. AT (collagenase digestion), UCB and BM (Ficoll gradient) were cultured (Dulbecco's modified Eagle's medium, low glucose, FBS) for 3 days. Detached adherent cells, at passage 4, were characterized as MSCs. Genetic stability was investigated by means of telomerase enzyme activity and karyotype. Hepatocyte differentiation protocol was performed with the use of Dulbecco's modified Eagle's medium, hepatocyte growth factor, basic fibroblast growth factor and nicotinamide (7 days); maturation medium (oncostatin, dexamethasone, insulin, transferrin and selenium) was added at 36 days. Hepatogenesis analyses were performed by use of morphology and albumin, AF, tyrosine-aminotransferase and glutamine synthetase gene expression and quantitative reverse transcription-polymerase chain reaction on days 9, 18, 25 and 36. Functionality was assessed through glycogen storage detection, indocyanine green absorption and transplantation procedure. U-MSCs and DHLCs were injected 48 h after induced fulminant hepatitis (intraperitoneal injection of carbon tetrachloride) in SCID/BALB-c mice. Histopathologic analyses were performed on days 7 and 15. Human origin included albumin and CK19 human markers. All MSCs differentiated into functional hepatocyte-like cells, stored glycogen and absorbed indocyanine green. AT-MSC DHLC gene expression was more consistent with a normal hepatogenic-differentiation profile. UCB-MSCs expanded weakly, impairing their use for the transplantation procedure. AT and BM U-MSCs and DHLCs regenerated liver injury equally. Regenerated hepatocytes exhibited human origin. AT might be the source and U-MSCS the stem cells useful for liver-regenerative therapy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
229857
Selenous acid, 99.999% trace metals basis
H2O3Se
211176
Selenous acid, 98%
H2O3Se