EMAIL THIS PAGE TO A FRIEND
EMBO molecular medicine
Microglial phagocytosis of living photoreceptors contributes to inherited retinal degeneration.
PMID 26139610
Abstract
Retinitis pigmentosa, caused predominantly by mutations in photoreceptor genes, currently lacks comprehensive treatment. We discover that retinal microglia contribute non-cell autonomously to rod photoreceptor degeneration by primary phagocytosis of living rods. Using rd10 mice, we found that the initiation of rod degeneration is accompanied by early infiltration of microglia, upregulation of phagocytic molecules in microglia, and presentation of "eat-me" signals on mutated rods. On live-cell imaging, infiltrating microglia interact dynamically with photoreceptors via motile processes and engage in rapid phagocytic engulfment of non-apoptotic rods. Microglial contribution to rod demise is evidenced by morphological and functional amelioration of photoreceptor degeneration following genetic ablation of retinal microglia. Molecular inhibition of microglial phagocytosis using the vitronectin receptor antagonist cRGD also improved morphological and functional parameters of degeneration. Our findings highlight primary microglial phagocytosis as a contributing mechanism underlying cell death in retinitis pigmentosa and implicate microglia as a potential cellular target for therapy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AB15282
Anti-Cone Arrestin Antibody, 100 µg, Anti-Cone Arrestin, Cat. No. AB15282, is a highly specific rabbit polyclonal antibody that targets Arrestin-C and has been tested in Immunohistochemistry (Paraffin) and Western Blotting.
05-719
Anti-Phosphatidylserine Antibody, clone 1H6, 200 µg, Anti-Phosphatidylserine Antibody, clone 1H6 is an antibody against Phosphatidylserine for use in FC, IH.
MAB5356
Anti-Rhodopsin Antibody, CT, last 9 amino acids, clone Rho 1D4, 100 µg, Anti-Rhodopsin Antibody, C-terminus, last 9 amino acids, clone Rho 1D4 detects level of Rhodopsin & has been published & validated for use in IC, IH, IP & WB.
GW10742A Anti-ARR3 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1405487
Anti-ARR3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB3501086
Anti-ARR3 antibody produced in rabbit, affinity isolated antibody
GW21443A Anti-EEA1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
AV30074
Anti-EEA1 antibody produced in rabbit, affinity isolated antibody
SAB2100649
Anti-EEA1 antibody produced in rabbit, affinity isolated antibody
SAB4300682
Anti-EEA1 antibody produced in rabbit, affinity isolated antibody
HPA038158
Anti-EEA1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA038159
Anti-EEA1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1306126
ANTI-HMFG(C-TERMINAL) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1406119
Anti-MFGE8 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA002807
Anti-MFGE8 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1408603
Anti-MFGE8 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
GW22359F Anti-PBR antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB2501221
Anti-Tspo (C-terminal) antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1405525
Anti-TSPO antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1411347
Anti-TSPO antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1402694
Monoclonal Anti-ARR3 antibody produced in mouse, clone 2D7, purified immunoglobulin, buffered aqueous solution
WH0000706M1
Monoclonal Anti-BZRP antibody produced in mouse, clone 3D8-B2, purified immunoglobulin, buffered aqueous solution
SAB1404579
Monoclonal Anti-EEA1, (C-terminal) antibody produced in mouse, clone 2G2, purified immunoglobulin, buffered aqueous solution
WH0004240M9
Monoclonal Anti-MFGE8 antibody produced in mouse, clone 3E9, purified immunoglobulin, buffered aqueous solution
SAB1404073
Monoclonal Anti-MFGE8 antibody produced in mouse, clone 1E7, purified immunoglobulin, buffered aqueous solution
T9778
Tropicamide, solid
C17H20N2O2