EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Interaction between mitsugumin 29 and TRPC3 participates in regulating Ca(2+) transients in skeletal muscle.
PMID 26141232
Abstract
Mitsugumin 29 (MG29) is related to the fatigue and aging processes of skeletal muscle. To examine the roles of MG29 in conjunction with its binding protein, the canonical-type transient receptor potential cation channel 3 (TRPC3), in skeletal muscle, the binding region of MG29 to TRPC3 was studied along with the functional relevance of the binding in mouse primary skeletal myotubes using co-immunoprecipitation assays and Ca(2+) imaging experiments. The N-terminus and the I-II loop of MG29 constitute the binding region for TRPC3. The myotubes that expressed the MG29 mutant missing the entire TRPC3-binding region showed a disrupted binding between endogenous MG29 and TRPC3 and a reduction in Ca(2+) transients in response to membrane depolarization without affecting ryanodine receptor 1 activity, the resting cytosolic Ca(2+) level, and the amount of releasable Ca(2+) from the sarcoplasmic reticulum. Among the proteins mediating Ca(2+) movements in skeletal muscle, TRPC4 expression was significantly decreased by the MG29 mutant. Therefore, MG29 could be a new factor for regulating Ca(2+) transients during skeletal muscle contraction possibly via a correlation with TRPC3 and TRPC4.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4502027
Anti-ATP2A1 antibody produced in rabbit, affinity isolated antibody
SAB2104862
Anti-ATP2A1, (N-terminal) antibody produced in rabbit, affinity isolated antibody
AV49532
Anti-JPH1 (AB1) antibody produced in rabbit, affinity isolated antibody
AV49533
Anti-JPH1 (AB2) antibody produced in rabbit, IgG fraction of antiserum
HPA009413
Anti-JPH1 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA008996
Anti-JPH1 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB3500328
Anti-JPH1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS4919
Anti-JPH2 (ab1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS4929
Anti-JPH2 (ab2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
AV51358
Anti-JPH2 antibody produced in rabbit, affinity isolated antibody
HPA052646
Anti-JPH2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2106850
Anti-JPH2 antibody produced in rabbit, affinity isolated antibody
SAB2900782
Anti-JPH2 antibody produced in rabbit, affinity isolated antibody
SAB2700849
Anti-QDPR antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA058951
Anti-QDPR antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
S6197
Anti-STIM1 (C-terminal) antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
S6072
Anti-STIM1 (N-terminal) antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB2501933 Anti-STIM1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA011088
Anti-STIM1 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA012123
Anti-STIM1 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB3500365
Anti-STIM1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3500069
Anti-SYPL2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2900851
Anti-SYPL2 antibody produced in rabbit, affinity isolated antibody
PRS3895
Anti-TRPC3 (ab1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS3905
Anti-TRPC3 (ab3) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
T5067
Anti-TRPC3 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
HPA037969
Anti-TRPC3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2103236
Anti-TRPC3, (N-terminal) antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB2501064
Anti-TRPC4 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2102582
Anti-TRPC4 antibody produced in rabbit, affinity isolated antibody, lyophilized powder