EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Habitual exercise training acts as a physiological stimulator for constant activation of lipolytic enzymes in rat primary white adipocytes.
PMID 26141235
Abstract
It is widely accepted that lipolysis in adipocytes are regulated through the enzymatic activation of both hormone-sensitive lipase (HSL) and adipose triglyceride lipase (ATGL) via their phosphorylation events. Accumulated evidence shows that habitual exercise training (HE) enhances the lipolytic response in primary white adipocytes with changes in the subcellular localization of lipolytic molecules. However, no study has focused on the effect that HE exerts on the phosphorylation of both HSL and ATGL in primary white adipocytes. It has been shown that the translocation of HSL from the cytosol to lipid droplet surfaces requires its phosphorylation at Ser-563. In primary white adipocytes obtained from HE rats, the level of HSL and ATGL proteins was higher than that in primary white adipocytes obtained from sedentary control (SC) rats. In HE rats, the level of phosphorylated ATGL and HSL was also significantly elevated compared with that in SC rats. These differences were confirmed by Phos-tag SDS-PAGE, a technique used to measure the amount of total phosphorylated proteins. Our results suggest that HE can consistently increase the activity of both lipases, thereby enhancing the lipolysis in white fat cells. Thus, HE helps in the prevention and treatment of obesity-related diseases by enhancing the lipolytic capacity.