EMAIL THIS PAGE TO A FRIEND
Atherosclerosis
Significant abnormalities of the HDL phosphosphingolipidome in type 1 diabetes despite normal HDL cholesterol concentration.
PMID 26142685
Abstract
Phospholipids and sphingolipids are major components of HDL. They play a critical role in HDL functionality and protective effects against atherosclerosis. As HDL are dysfunctional in type 1 diabetic patients, we ascertained whether they presented abnormalities in their phospholipid and sphingolipid profile, despite normal HDL cholesterol concentration. Using liquid chromatography-tandem mass spectrometry, we quantified the main species of phosphatidylcholines, sphingomyelins, lysophophatidylcholines, phosphatidylethanolamines, phosphatidylinositols, ceramides, plasmalogens and sphingosines 1-phosphate in the HDL2 and HDL3 from 54 type 1 diabetic patients and 50 controls. Serum HDL cholesterol was similar in the 2 groups of subjects. When data were expressed relative to the total amount of phospholipids and sphingolipids, sphingosines-1-phosphate (S1P) were 11.7% (NS) and 14.4% (pxa0=xa00.0062) lower in HDL2 and HDL3, respectively, from type 1 diabetic patients than from controls. Ceramides were 23% (pxa0=xa00.005) and 24% (borderline significance) lower in HDL2 and HDL3, respectively. The concentration of apolipoprotein M, the carrier of S1P, was similar in patients and controls. In type 1 diabetic patients compared to controls, the concentration of d18:1-S1P, the main S1P species, was decreased in total plasma (-17.0%, pxa0
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
59304
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
C3H8O
40301
2-Propanol, semiconductor grade VLSI PURANAL (Honeywell 17604)
C3H8O
40219
2-Propanol, semiconductor grade PURANAL (Honeywell 17829)
C3H8O
40279
2-Propanol, semiconductor grade MOS PURANAL (Honeywell 17930)
C3H8O
270490
2-Propanol, 99.5%, HPLC grade
C3H8O
15-2360
2-Propanol, for HPLC
C3H8O
733458
2-Propanol, electronic grade, 99.999% trace metals basis
C3H8O
C7559
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
CHCl3
372978
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
CHCl3
05-3433
Chloroform, for residue analysis, suitable for 5000 per JIS
CHCl3
05-3450
Chloroform, for HPLC
CHCl3
270989
Ethyl acetate, anhydrous, 99.8%
C4H8O2
437549
Ethyl acetate, ACS reagent, ≥99.5%
C4H8O2
324027
Ethyl acetate, PRA grade, ≥99.5%
C4H8O2
270520
Ethyl acetate, HPLC grade, 99.8%
C4H8O2
W241407
Ethyl acetate, ≥99%, FCC, FG
C4H8O2
09-0703
Ethyl acetate, for residue analysis, suitable for 5000 per JIS
C4H8O2
09-0730
Ethyl acetate, for HPLC
C4H8O2
W241415
Ethyl acetate, natural, ≥99%, FCC, FG
C4H8O2
537446
Ethyl acetate, ReagentPlus®, ≥99.8%
C4H8O2
L092004
Ethyl acetate
C4H8O2
L092003
Ethyl acetate
C4H8O2
W292907
Isopropyl alcohol, ≥99.7%, FCC, FG
C3H8O