EMAIL THIS PAGE TO A FRIEND
Academic radiology
Screening Breast MRI in Patients Previously Treated for Breast Cancer: Diagnostic Yield for Cancer and Abnormal Interpretation Rate.
PMID 26142951
Abstract
To determine the cancer detection rate and abnormal interpretation rate of screening breast magnetic resonance imaging (MRI) in previously treated breast cancer patients. Institutional review board-approved retrospective review of the breast MRI database from 2009 to 2011 identified a total of 3297 screening examinations. After excluding genetic mutation carriers, untested first-degree relatives of known mutation carriers, and patients with a history of chest irradiation, there were 1194 (36.2%) examinations in 691 patients previously treated for breast cancer. MRI reports were reviewed to determine MRI findings and breast imaging reporting and data system (BI-RADS) assessments. The longitudinal medical record was reviewed to determine patient demographics and outcomes of imaging surveillance and biopsy. Mean patient age at initial cancer diagnosis was 46.1 years, and mean patient age during the study interval was 52 years. Cancer detection rate was 10 per 1000 (1%; 95% confidence interval [CI], 0.5%-1.8%]; 12 of 1194 examinations). Overall 10.7% (128 of 1194) of examinations received an abnormal interpretation, including 5.4% (65 of 1194) BI-RADS 4 or 5 and 5.3% (63 of 1194) BI-RADS 3 assessments with a 9.4% positive predictive value (PPV1; 12 of 128 examinations) and a 17.9% PPV3 (12 malignancies per 67 biopsies). Screening breast MRI in women previously treated for breast cancer detected cancer in 1.0% of examinations, with a 10.7% abnormal interpretation rate, and a PPV for malignancy of 17.9%.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A6508
L-Aspartic acid β-hydroxamate, serine racemase inhibitor
C4H8N2O4