EMAIL THIS PAGE TO A FRIEND
Scientific reports
Formation and manipulation of cell spheroids using a density adjusted PEG/DEX aqueous two phase system.
PMID 26144552
Abstract
Various spheroid formation techniques have been widely developed for efficient and reliable 3-D cell culture research. Although those efforts improved many aspects of spheroid generation, the procedures became complex and also required unusual laboratory equipment. Many recent techniques still involve laborious pipetting steps for spheroid manipulation such as collection, distribution and reseeding. In this report, we used a density-controlled polyethylene glycol and dextran aqueous two phase system to generate spheroids that are both consistent in size and precisely size-controllable. Moreover, by adding a few drops of fresh medium to the wells the contain spheroids, they can be simply settled and attached to the culture surface due to reduced densities of the phases. This unique attribute of the technique significantly reduces the numerous pipetting steps of spheroid manipulation to a single pipetting; therefore, the errors from those steps are eliminated and the reliability and efficiency of a research can be maximized.