EMAIL THIS PAGE TO A FRIEND
Placenta
Transcription factors E2F1 and E2F3 are expressed in placenta but do not regulate MMP14.
PMID 26145228
Abstract
Preeclampsia is a serious complication of pregnancy for which there are no efficacious medical treatments. Soluble endoglin is as an anti-angiogenic factor that contributes to the pathogenesis of the disease, however little is known about its molecular regulation in placenta. Recent data has demonstrated E2F transcription factors directly regulate MMPs in metastatic disease. Of particular interest was the capacity of E2F1 and E2F3 to up-regulate MMP14, a protease that cleaves and releases soluble endoglin from placenta. The aim of this study was to characterize E2F1 and E2F3 in preeclamptic placenta and assess whether silencing affects soluble endoglin release. E2F1 and E2F3 mRNA, protein expression and localization were assessed in severe early onset preeclamptic and preterm control placentas (delivered <34 weeks gestation). E2F siRNA was administered to primary trophoblast and primary endothelial cells and effect on MMP14 mRNA expression and soluble endoglin secretion assessed. E2F1 and E2F3 were localized to the syncytiotrophoblast. E2F1 was significantly down regulated in severe preeclamptic placentas, while E2F3 was unchanged. Silencing E2F1 did not decrease MMP14 expression in primary trophoblast or endothelial cells. However, E2F1 silencing resulted in a significant increase in soluble endoglin secretion from both cell types, and silencing of E2F3 also significantly increased soluble endoglin release from primary trophoblast. This study demonstrates that E2F1 and E2F3 are present within the syncytiotrophoblast of placenta and that E2F1 is reduced in preeclampsia. Although silencing of either E2F1 or E2F3 does not alter MMP14 expression, both appear to regulate soluble endoglin release.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV04006
Anti-E2F1 (AB1) antibody produced in rabbit, affinity isolated antibody
SAB2502110
Anti-E2F1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA008003
Anti-E2F1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
E9026 Anti-E2F1 antibody produced in rabbit, ~0.8 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB2103144
Anti-E2F1, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB4500682
Anti-E2F1, C-Terminal antibody produced in rabbit, affinity isolated antibody
AV38089
Anti-E2F3 antibody produced in rabbit, affinity isolated antibody
RAB0171
Human Endoglin ELISA Kit, for serum, plasma, cell culture supernatants and urine
EHU070981 MISSION® esiRNA, esiRNA human E2F1 (esiRNA1)
EMU075181 MISSION® esiRNA, esiRNA targeting mouse E2f1 (esiRNA1)
WH0001869M1
Monoclonal Anti-E2F1 antibody produced in mouse, clone 2E10, purified immunoglobulin, buffered aqueous solution
E8901 Monoclonal Anti-E2F1 antibody produced in mouse, ~0.5 mg/mL, clone KH20, purified immunoglobulin, buffered aqueous solution
WH0001871M1
Monoclonal Anti-E2F3 antibody produced in mouse, clone 5F7, purified immunoglobulin, buffered aqueous solution
E8651 Monoclonal Anti-E2F3 antibody produced in mouse, ~2 mg/mL, clone PG30, purified immunoglobulin, buffered aqueous solution
SAB1412705
Monoclonal Anti-E2F3 antibody produced in mouse, clone 3C11, purified immunoglobulin, 1 mg/mL
RAB0782 Mouse Eng / Endoglin ELISA Kit
PLA0139
Rabbit anti-E2F1 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.