PloS one
A Novel Insertion Variant of CRYGD Is Associated with Congenital Nuclear Cataract in a Chinese Family.
PMID 26147294
Abstract
To investigate a novel insertion variant of CRYGD identified in a Chinese family with nuclear congenital cataract. A Chinese family with congenital nuclear cataract was recruited for the mutational screening of candidate genes by direct sequencing. Recombinant N-terminal Myc tagged wildtype or mutant CRYGD was expressed in HEK293T cells. The expression pattern, protein solubility and subcellular distribution were analyzed by western blotting and immunofluorescence. A novel insertion variant, c.451_452insGACT, in CRYGD was identified in the patients. It causes a frameshift and a premature termination of the polypeptide to become Y151*. A significantly reduced solubility was observed for this mutant. Unlike wildtype CRYGD, which existed mainly in the cytoplasm, Y151* was mis-located in the nucleus. We have identified a novel mutation, c.451_452insGACT, in CRYGD, which is associated with nuclear cataract. This is the first insertion mutation of CRYGD found to cause autosomal dominant congenital cataract. The mutant protein, with loss of solubility and localization to the nucleus, is hypothesized to be the major cause of cataract in these patients.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB3701028 Anti-Mouse IgG (Fc specific), F(ab′)2 fragment, highly cross adsorbed-Fluorescein antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3701011 Anti-Mouse IgG (Fc specific), F(ab′)2 fragment, highly cross adsorbed-Rhodamine antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3701033 Anti-Mouse IgG (Fc specific), F(ab′)2 fragment, highly cross adsorbed-Texas Red™ antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB4600013
Anti-Mouse IgG (H+L), CF 350 antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600019
Anti-Mouse IgG (H+L), CF 405M antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600023
Anti-Mouse IgG (H+L), CF 405S antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600042
Anti-Mouse IgG (H+L), CF 488A antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600066
Anti-Mouse IgG (H+L), CF 555 antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600082
Anti-Mouse IgG (H+L), CF 568 antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600105
Anti-Mouse IgG (H+L), CF 594 antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600138
Anti-Mouse IgG (H+L), CF 633 antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600161
Anti-Mouse IgG (H+L), CF 640R antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600182
Anti-Mouse IgG (H+L), CF 647 antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600191
Anti-Mouse IgG (H+L), CF 660C antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600194
Anti-Mouse IgG (H+L), CF 660R antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600211
Anti-Mouse IgG (H+L), CF 750 antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB4600222
Anti-Mouse IgG (H+L), F(ab′)2 fragment, CF350 antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4600405
Anti-Mouse IgG (H+L), F(ab′)2 fragment, CF350 antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4600387
Anti-Mouse IgG (H+L), F(ab′)2 fragment, CF488A antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4600388
Anti-Mouse IgG (H+L), F(ab′)2 fragment, CF488A antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4600299
Anti-Mouse IgG (H+L), F(ab′)2 fragment, CF555 antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4600390
Anti-Mouse IgG (H+L), F(ab′)2 fragment, CF555 antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4600401
Anti-Mouse IgG (H+L), F(ab′)2 fragment, CF568 antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4600321
Anti-Mouse IgG (H+L), F(ab′)2 fragment, CF594 antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4600402
Anti-Mouse IgG (H+L), F(ab′)2 fragment, CF594 antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4600403
Anti-Mouse IgG (H+L), F(ab′)2 fragment, CF633 antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4600396
Anti-Mouse IgG (H+L), F(ab′)2 fragment, CF640R antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4600397
Anti-Mouse IgG (H+L), F(ab′)2 fragment, CF640R antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4600351
Anti-Mouse IgG (H+L), F(ab′)2 fragment, CF647 antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4600392
Anti-Mouse IgG (H+L), F(ab′)2 fragment, CF647 antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution