PloS one
Potent and Selective Triazole-Based Inhibitors of the Hypoxia-Inducible Factor Prolyl-Hydroxylases with Activity in the Murine Brain.
PMID 26147748
Abstract
As part of the cellular adaptation to limiting oxygen availability in animals, the expression of a large set of genes is activated by the upregulation of the hypoxia-inducible transcription factors (HIFs). Therapeutic activation of the natural human hypoxic response can be achieved by the inhibition of the hypoxia sensors for the HIF system, i.e. the HIF prolyl-hydroxylases (PHDs). Here, we report studies on tricyclic triazole-containing compounds as potent and selective PHD inhibitors which compete with the 2-oxoglutarate co-substrate. One compound (IOX4) induces HIFα in cells and in wildtype mice with marked induction in the brain tissue, revealing that it is useful for studies aimed at validating the upregulation of HIF for treatment of cerebral diseases including stroke.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
07-1585
Anti-HIF-1-alpha Antibody, hydroxyproline (Pro402), 100 µg, Use Anti-HIF-1-alpha Antibody, hydroxyproline (Pro402) (Rabbit Polyclonal Antibody) validated in WB to detect HIF-1-alpha also known as Hypoxia-inducible factor 1-alpha, HIF1-alpha.
C4491 Anti-Calcium-Sensing Receptor antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1405561
Anti-CASR antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA039686
Anti-CASR antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV34608
Anti-EGLN1 antibody produced in rabbit, IgG fraction of antiserum
HPA022129
Anti-EGLN1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV39316
Anti-HIF1AN (AB1) antibody produced in rabbit, affinity isolated antibody
AV32904
Anti-HIF1AN (AB2) antibody produced in rabbit, affinity isolated antibody
AV32903
Anti-HIF1AN (AB3) antibody produced in rabbit, IgG fraction of antiserum
SAB2101040
Anti-HIF1AN antibody produced in rabbit, affinity isolated antibody
86062703
Hep 3B Cell Line human, liver from human
WH0000846M1 Monoclonal Anti-CASR antibody produced in mouse, clone 3F12, purified immunoglobulin, buffered aqueous solution
WH0055662M1
Monoclonal Anti-HIF1AN antibody produced in mouse, clone 1D8, purified immunoglobulin, buffered aqueous solution