PloS one
Vascular Adventitia Calcification and Its Underlying Mechanism.
PMID 26148272
Abstract
Previous research on vascular calcification has mainly focused on the vascular intima and media. However, we show here that vascular calcification may also occur in the adventitia. The purpose of this work is to help elucidate the pathogenic mechanisms underlying vascular calcification. The calcified lesions were examined by Von Kossa staining in ApoE-/- mice which were fed high fat diets (HFD) for 48 weeks and human subjects aged 60 years and older that had died of coronary heart disease, heart failure or acute renal failure. Explant cultured fibroblasts and smooth muscle cells (SMCs)were obtained from rat adventitia and media, respectively. After calcification induction, cells were collected for Alizarin Red S staining. Calcified lesions were observed in the aorta adventitia and coronary artery adventitia of ApoE-/-mice, as well as in the aorta adventitia of human subjects examined. Explant culture of fibroblasts, the primary cell type comprising the adventitia, was successfully induced for calcification after incubation with TGF-β1 (20 ng/ml) + mineralization media for 4 days, and the phenotype conversion vascular adventitia fibroblasts into myofibroblasts was identified. Culture of SMCs, which comprise only a small percentage of all cells in the adventitia, in calcifying medium for 14 days resulted in significant calcification.Vascular calcification can occur in the adventitia. Adventitia calcification may arise from the fibroblasts which were transformed into myofibroblasts or smooth muscle cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV48225
Anti-VIM (AB1) antibody produced in rabbit, affinity isolated antibody
AV48226
Anti-VIM (AB2) antibody produced in rabbit, affinity isolated antibody
SAB1406585
Anti-VIM antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA001762
Anti-VIM antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
V4630 Anti-Vimentin antibody produced in goat, whole antiserum
SAB4300676
Anti-Vimentin antibody produced in rabbit, affinity isolated antibody
SAB4503081
Anti-Vimentin antibody produced in rabbit, affinity isolated antibody
SAB4503083
Anti-Vimentin, C-Terminal antibody produced in rabbit, affinity isolated antibody
WH0007431M1
Monoclonal Anti-VIM antibody produced in mouse, clone 3E9, purified immunoglobulin, buffered aqueous solution
SAB4700524 Monoclonal Anti-VIM antibody produced in mouse, clone VI-RE/1, purified immunoglobulin, buffered aqueous solution
SAB4100612
Monoclonal Anti-VIM antibody produced in mouse, clone VIM-A604, culture supernatant
AMAB90516
Monoclonal Anti-VIM antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0157, purified immunoglobulin, buffered aqueous glycerol solution
V6389
Monoclonal Anti-Vimentin antibody produced in mouse, ~1 mg/mL, clone V9, purified immunoglobulin, buffered aqueous solution
V5255
Monoclonal Anti-Vimentin antibody produced in mouse, clone VIM-13.2, ascites fluid
V2258
Monoclonal Anti-Vimentin antibody produced in mouse, clone LN-6, ascites fluid
PLA0199
Rabbit anti-Vimentin Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.