EMAIL THIS PAGE TO A FRIEND
Journal of bacteriology
Energy Conservation Associated with Ethanol Formation from H2 and CO2 in Clostridium autoethanogenum Involving Electron Bifurcation.
PMID 26148714
Abstract
Most acetogens can reduce CO2 with H2 to acetic acid via the Wood-Ljungdahl pathway, in which the ATP required for formate activation is regenerated in the acetate kinase reaction. However, a few acetogens, such as Clostridium autoethanogenum, Clostridium ljungdahlii, and Clostridium ragsdalei, also form large amounts of ethanol from CO2 and H2. How these anaerobes with a growth pH optimum near 5 conserve energy has remained elusive. We investigated this question by determining the specific activities and cofactor specificities of all relevant oxidoreductases in cell extracts of H2/CO2-grown C. autoethanogenum. The activity studies were backed up by transcriptional and mutational analyses. Most notably, despite the presence of six hydrogenase systems of various types encoded in the genome, the cells appear to contain only one active hydrogenase. The active [FeFe]-hydrogenase is electron bifurcating, with ferredoxin and NADP as the two electron acceptors. Consistently, most of the other active oxidoreductases rely on either reduced ferredoxin and/or NADPH as the electron donor. An exception is ethanol dehydrogenase, which was found to be NAD specific. Methylenetetrahydrofolate reductase activity could only be demonstrated with artificial electron donors. Key to the understanding of this energy metabolism is the presence of membrane-associated reduced ferredoxin:NAD(+) oxidoreductase (Rnf), of electron-bifurcating and ferredoxin-dependent transhydrogenase (Nfn), and of acetaldehyde:ferredoxin oxidoreductase, which is present with very high specific activities in H2/CO2-grown cells. Based on these findings and on thermodynamic considerations, we propose metabolic schemes that allow, depending on the H2 partial pressure, the chemiosmotic synthesis of 0.14 to 1.5 mol ATP per mol ethanol synthesized from CO2 and H2. Ethanol formation from syngas (H2, CO, and CO2) and from H2 and CO2 that is catalyzed by bacteria is presently a much-discussed process for sustainable production of biofuels. Although the process is already in use, its biochemistry is only incompletely understood. The most pertinent question is how the bacteria conserve energy for growth during ethanol formation from H2 and CO2, considering that acetyl coenzyme A (acetyl-CoA), is an intermediate. Can reduction of the activated acetic acid to ethanol with H2 be coupled with the phosphorylation of ADP? Evidence is presented that this is indeed possible, via both substrate-level phosphorylation and electron transport phosphorylation. In the case of substrate-level phosphorylation, acetyl-CoA reduction to ethanol proceeds via free acetic acid involving acetaldehyde:ferredoxin oxidoreductase (carboxylate reductase).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R4500
(−)-Riboflavin, from Eremothecium ashbyii, ≥98%
C17H20N4O6
R7649
(−)-Riboflavin, meets USP testing specifications
C17H20N4O6
R0508
(−)-Riboflavin, acrylamide photopolymerization tested
C17H20N4O6
R9504
(−)-Riboflavin, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
C17H20N4O6
402788
Acetaldehyde, ACS reagent, ≥99.5%
C2H4O
W200336
Acetaldehyde, natural, ≥99%, FG
C2H4O
W200301
Acetaldehyde, ≥99%, meets FCC analytical specification
C2H4O
W200360
Acetaldehyde, ≥99%, meets FCC analytical specification
C2H4O
110078
Acetaldehyde, ReagentPlus®, 99%
C2H4O
W200320
Acetaldehyde, ≥99%, FCC, stabilized
C2H4O
W200379
Acetaldehyde solution, 40 wt. % in H2O
C2H4O
W200387
Acetaldehyde solution, 40 wt. % in isopropanol
C2H4O
W200344
Acetaldehyde solution, natural, 50 wt. % ethanol, FG
C2H4O
W200328
Acetaldehyde solution, 50 wt. % in ethanol
C2H4O
719099
Acetaldehyde solution, 5 M in THF
C2H4O
W200300
Acetaldehyde solution, natural, 50 wt. % in ethanol
C2H4O
N4126
Nicotinic acid, ≥98%
C6H5NO2
72309
Nicotinic acid, ≥99.5% (HPLC)
C6H5NO2
N0761
Nicotinic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
C6H5NO2
N5410
Nicotinic acid, meets USP testing specifications
C6H5NO2
P9755
Pyridoxine hydrochloride, ≥98% (HPLC)
C8H11NO3 · HCl
P4722
Pyridoxine hydrochloride, meets USP testing specifications
C8H11NO3 · HCl
P6280
Pyridoxine hydrochloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture
C8H11NO3 · HCl
C8754
Thiamine pyrophosphate, ≥95%
C12H19ClN4O7P2S