EMAIL THIS PAGE TO A FRIEND
Experimental eye research
Deficiency of CC chemokine ligand 2 and decay-accelerating factor causes retinal degeneration in mice.
PMID 26149093
Abstract
CC chemokine ligand 2 (CCL2) recruits macrophages to reduce inflammatory responses. Decay-accelerating factor (DAF) is a membrane regulator of the classical and alternative pathways of complement activation. In view of the link between complement genes and retinal diseases, we evaluated the retinal phenotype of C57BL/6J mice and mice lacking Ccl2 and/or Daf1 at 12 months of age, using scanning laser ophthalmoscopic imaging, electroretinography (ERG), histology, immunohistochemistry, and terminal deoxynucleotidyl transferase dUTP nick end labeling (TUNEL) analysis. In comparison to C57BL/6J mice, mutant mice had an increased number of autofluorescent foci, with the greatest number in the Ccl2(-/-)/Daf1(-/-) retina. ERG amplitudes in Ccl2(-/-)/Daf1(-/-), Ccl2(-/-) and Daf1(-/-) mice were reduced, with the greatest reduction in Ccl2(-/-)/Daf1(-/-) mice. TUNEL-positive cells were not seen in C57BL/6J retina, but were prevalent in the outer and inner nuclear layers of Ccl2(-/-)Daf1(-/-) mice and were present at reduced density in Ccl2(-/-) or Daf1(-/-) mice. Cell loss was most pronounced in the outer and inner nuclear layers of Ccl2(-/-)/Daf1(-/-) mice. The levels of the endoplasmic reticulum chaperone GPR78 and transcription factor ATF4 were significantly increased in the Ccl2(-/-)/Daf1(-/-) retina. In comparison to the C57BL/6J retina, the phosphorylation of NF-κB p65, p38, ERK and JNK was significantly upregulated while SIRT1 was significantly downregulated in the Ccl2(-/-)/Daf1(-/-) retina. Our results suggest that loss of Ccl2 and Daf1 causes retinal neuronal death and degeneration which is related to increased endoplasmic reticulum stress, oxidative stress and inflammation.