EMAIL THIS PAGE TO A FRIEND
Kidney international
Urinary uromodulin, kidney function, and cardiovascular disease in elderly adults.
PMID 26154925
Abstract
Urinary uromodulin (uUMOD) is the most common secreted tubular protein in healthy adults. However, the relationship between uUMOD and clinical outcomes is still unclear. Here we measured uUMOD in 192 participants of the Cardiovascular Health Study with over a 30% decline in estimated glomerular filtration rate (eGFR) over 9 years, 54 with incident end-stage renal disease (ESRD), and in a random subcohort of 958 participants. The association of uUMOD with eGFR decline was evaluated using logistic regression and with incident ESRD, cardiovascular disease, heart failure, and mortality using Cox proportional regression. Mean age was 78 years and median uUMOD was 25.8 μg/ml. In a case-control study evaluating eGFR decline (192 cases and 231 controls), each 1-s.d. higher uUMOD was associated with a 23% lower odds of eGFR decline (odds ratio 0.77 (95% CI 0.62-0.96)) and a 10% lower risk of mortality (hazard ratio 0.90 (95% CI 0.83-0.98)) after adjusting for demographics, eGFR, albumin/creatinine ratio, and other risk factors. There was no risk association of uUMOD with ESRD, cardiovascular disease, or heart failure after multivariable adjustment. Thus, low uUMOD levels may identify persons at risk of progressive kidney disease and mortality above and beyond established markers of kidney disease, namely eGFR and the albumin/creatinine ratio. Future studies need to confirm these results and evaluate whether uUMOD is a marker of tubular health and/or whether it plays a causal role in preserving kidney function.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
RAB0751 Human UMOD / Uromodulin ELISA Kit