EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Preparation and Characterization of Dictyophora indusiata Polysaccharide-Zinc Complex and Its Augmented Antiproliferative Activity on Human Cancer Cells.
PMID 26155804
Abstract
Dictyophora indusiata polysaccharide (DP1) was successfully chelated with zinc chloride to achieve its enhanced antiproliferative activity. The obtained DP1-Zn complex showed significant antiproliferative activity (18.1 ± 2.84% viability of MCF-7 cells at 250 μg/mL) on a group of human cancer cell lines through induction of apoptosis. The pro-apoptotic action of DP1-Zn was confirmed by morphological changes including chromatin condensation, DNA breakage, and S phase cell cycle arrest in human breast adenocarcinoma cells (MCF-7). The DP1-Zn-induced apoptotic pathways were characterized by the activation of caspases-3, -8, and -9, mitochondrial dysfunction, and reactive oxygen species (ROS) overproduction (305 ± 7.06% production of control at 250 μg/mL). This study suggested that DP1-Zn can be developed as a candidate for cancer treatment and prevention, especially human breast adenocarcinoma.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A5845
N-Acetyl-Leu-Glu-His-Asp-7-amido-4-trifluoromethylcoumarin, ≥95% (HPLC), powder
C33H38O11N7F3