EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Pituitary Adenylate Cyclase-activating Polypeptide (PACAP) Targets Down Syndrome Candidate Region 1 (DSCR1/RCAN1) to control Neuronal Differentiation.
PMID 26157140
Abstract
Pituitary adenylate cyclase-activating peptide (PACAP) is a neurotrophic peptide involved in a wide range of nervous functions, including development, differentiation, and survival, and various aspects of learning and memory. Here we report that PACAP induces the expression of regulator of calcineurin 1 (RCAN1, also known as DSCR1), which is abnormally expressed in the brains of Down syndrome patients. Increased RCAN1 expression is accompanied by activation of the PKA-cAMP response element-binding protein pathways. EMSA and ChIP analyses demonstrate the presence of a functional cAMP response element in the RCAN1 promoter. Moreover, we show that PACAP-dependent neuronal differentiation is significantly disturbed by improper RCAN1 expression. Our data provide the first evidence of RCAN1, a Down syndrome-related gene, as a novel target for control of the neurotrophic function of PACAP.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D6694
Anti-DSCR1 (C-terminal) antibody produced in rabbit, ~1.5 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB1305103 ANTI-DSCR1 (CENTER) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1405729
Anti-DSCR1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1305104 ANTI-DSCR1(C-TERMINAL) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB2501991
Anti-RCAN1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2101967
Anti-RCAN1 antibody produced in rabbit, affinity isolated antibody
SAB4300513
Anti-SYN1 (Ab-9) antibody produced in rabbit, affinity isolated antibody
SAB1412138
ANTI-SYN1 antibody produced in mouse, clone 3F12, purified immunoglobulin, buffered aqueous solution
SAB1412425
ANTI-SYN1 antibody produced in mouse, clone 3H6, purified immunoglobulin, buffered aqueous solution
SAB1412529
ANTI-SYN1 antibody produced in mouse, clone 3E3, purified immunoglobulin, buffered aqueous solution
SAB1412557
ANTI-SYN1 antibody produced in mouse, clone 1D10, purified immunoglobulin, buffered aqueous solution
HPA000397
Anti-SYN1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
S193 Anti-Synapsin I antibody produced in rabbit, affinity isolated antibody, lyophilized powder
WH0001827M1 Monoclonal Anti-DSCR1 antibody produced in mouse, clone 1G7, ascites fluid, buffered aqueous solution
WH0001827M3
Monoclonal Anti-DSCR1 antibody produced in mouse, clone 1B1, purified immunoglobulin, buffered aqueous solution
WH0006853M7
Monoclonal Anti-SYN1 antibody produced in mouse, clone 4H1, purified immunoglobulin, buffered aqueous solution
SAB1404432
Monoclonal Anti-SYN1 antibody produced in mouse, clone 4F6, purified immunoglobulin, buffered aqueous solution