EMAIL THIS PAGE TO A FRIEND
Chinese journal of cancer research = Chung-kuo yen cheng yen chiu
microRNA-218 promotes gemcitabine sensitivity in human pancreatic cancer cells by regulating HMGB1 expression.
PMID 26157323
Abstract
The purpose of this study was to examine the effect of gemcitabine (GEM) on microRNA-218 (miR-218) expression in human pancreatic cancer cells. Quantitative reverse transcription polymerase chain reaction (qRT-PCR) was performed to examine the differences in miR-218 expression between the GEM-sensitive BxPC-3 pancreatic cancer cells and GEM-resistant PANC-1 cells. The effect of GEM on the expression of miR-218 in PANC-1 cells was also investigated. PANC-1 cells were transfected either with HMGB1 siRNA to knock down the expression of HMGB1 or with the recombinant HMGB1 expression vector (pcDNA3.1-HMGB1) to overexpress HMGB1. The effect of ectopic expression of HMGB1 on the apoptosis of miR-218-transfected and GEM-treated PANC-1 cells was examined by flow cytometric analysis. The miR-218 expression level was lower in GEM-resistant PANC-1 cells compared to GEM-sensitive BxPC-3 cells (P<0.05). The percentage of apoptotic PANC-1 cells was significantly increased in the miR-218 mimic + GEM group compared to the mimic ctrl + GEM group and the normal control group (P<0.01). The HMGB1 expression level was markedly decreased in PANC-1 cells transfected with HMGB1 siRNA but was significantly increased in PANC-1 cells transfected with the recombinant HMGB1 expression vector, pcDNA3.1-HMGB1 (P<0.01). The proportion of apoptotic PANC-1 cells was significantly lower in the miR-218 mimic + GEM + pcDNA3.1-HMGB1 group compared to the miR-218 mimic + GEM + HMGB1 siRNA group (P<0.01). The expression level of miR-218 was downregulated in the GEM-resistant cell line. miR-218 promoted the sensitivity of PANC-1 cells to GEM, which was achieved mainly through regulating the expression of HMGB1 in PANC-1 cells.