EMAIL THIS PAGE TO A FRIEND
BMC infectious diseases
Fecal microbiota transplantation restores dysbiosis in patients with methicillin resistant Staphylococcus aureus enterocolitis.
PMID 26159166
Abstract
Nosocomial Methicillin-resistant Staphylococcus aureus (MRSA) enteritis is rare but can be fatal unless it is detected at an early stage and treated effectively. Dysbiosis of the gut is one of the leading reasons of MRSA enteritis. Fecal microbiota transplantation (FMT) is a burgeoning treatment to rectify this imbalance. But the impact of FMT on MRSA enterocoitis is still unknown yet. A total of 5 patients diagnosed as MRSA enteritis during the early postoperative period were given vancomycin 2 g/day for 3 days and FMT for three continuous days as a standard treatment. There was a 100% clinical response rate that all the symptoms resulting from MRSA enterocolitis disappeared and MRSA in the feces eliminated clearly. The microbiota profile in feces of the patients also regained balance. FMT can be a preferential measure to restore the dysbiosis caused by MSRA enterocolitis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O104
Omeprazole, solid
C17H19N3O3S