EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
XRN2 is required for the degradation of target RNAs by RNase H1-dependent antisense oligonucleotides.
PMID 26159921
Abstract
Antisense oligonucleotides (ASOs) can suppress the expression of a target gene by cleaving pre-mRNA and/or mature mRNA via RNase H1. Following the initial endonucleolytic cleavage by RNase H1, the target RNAs are degraded by a mechanism that is poorly understood. To better understand this degradation pathway, we depleted the expression of two major 5' to 3' exoribonucleases (XRNs), named XRN1 and XRN2, and analyzed the levels of 3' fragments of the target RNAs in vitro. We found that the 3' fragments of target pre-mRNA generated by ASO were almost completely degraded from their 5' ends by nuclear XRN2 after RNase H1-mediated cleavage, whereas the 3' fragments of mature mRNA were partially degraded by XRN2. In contrast to ASO, small interference RNA (siRNA) could reduce the expression level of only mature mRNA, and the 3' fragment was degraded by cytoplasmic XRN1. Our findings indicate that the RNAs targeted by RNase H1-dependent ASO are rapidly degraded in the nucleus, contrary to the cytoplasmic degradation pathway mediated by siRNA.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1412938
Anti-RNASEH1 antibody produced in mouse, purified immunoglobulin, 0.5 mg/mL
SAB2102013
Anti-RNASEH1 antibody produced in rabbit, affinity isolated antibody
SAB2701325
Anti-RNASEH1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2102729
Anti-XRN1 antibody produced in rabbit, affinity isolated antibody
SAB4200028 Anti-XRN1 antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB1401727
Anti-XRN1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA035005 Anti-XRN1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA035006 Anti-XRN1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1304705 ANTI-XRN2 (N-TERM) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
HPA047118
Anti-XRN2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
EHU106741 MISSION® esiRNA, esiRNA human RNASEH1 (esiRNA1)
EHU043071 MISSION® esiRNA, esiRNA human XRN1 (esiRNA1)
EMU023091 MISSION® esiRNA, esiRNA targeting mouse Rnaseh1 (esiRNA1)
EHU049051 MISSION® esiRNA, esiRNA human XRN2 (esiRNA1)
EMU090681 MISSION® esiRNA, esiRNA targeting mouse Xrn1 (esiRNA1)
EMU085551 MISSION® esiRNA, esiRNA targeting mouse Xrn2 (esiRNA1)
WH0246243M1
Monoclonal Anti-RNASEH1 antibody produced in mouse, clone 5D10, purified immunoglobulin, buffered aqueous solution
PLA0105
Rabbit anti-XRN1 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.