EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
The Role of the Pleckstrin Homology Domain-containing Protein CKIP-1 in Activation of p21-activated Kinase 1 (PAK1).
PMID 26160174
Abstract
Upon growth factor stimulation, PAK1 is recruited to the plasma membrane and activated by a mechanism that requires its phosphorylation at Ser-223 by the protein kinase CK2. However, the upstream signaling molecules that regulate this phosphorylation event are not clearly defined. Here, we demonstrate a major role of the CK2α-interacting protein CKIP-1 in activation of PAK1. CK2α, CKIP-1, and PAK1 are translocated to membrane ruffles in response to the epidermal growth factor (EGF), where CKIP-1 mediates the interaction between CK2α and PAK1 in a PI3K-dependent manner. Consistently, PAK1 mediates phosphorylation and modulation of the activity of p41-Arc, one of its plasma membrane substrate, in a fashion that requires PI3K and CKIP-1. Moreover, CKIP-1 knockdown or PI3K inhibition suppresses PAK1-mediated cell migration and invasion, demonstrating the physiological significance of the PI3K-CKIP-1-CK2-PAK1 signaling pathway. Taken together, these findings identify a novel mechanism for the activation of PAK1 at the plasma membrane, which is critical for cell migration and invasion.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1306535
ANTI-CKIP-1(N-TERMINAL) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
AV53595
Anti-CSNK2A2 (AB1) antibody produced in rabbit, affinity isolated antibody
SAB1410253
Anti-CSNK2A2 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4300427
Anti-PAK1 (Ab-212) antibody produced in rabbit, affinity isolated antibody
SAB2500741
Anti-PAK1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1406219
Anti-PAK1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA003565
Anti-PAK1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4502062
Anti-PAK1 antibody produced in rabbit, affinity isolated antibody
SAB4502064
Anti-PAK1 antibody produced in rabbit, affinity isolated antibody
SAB4502065
Anti-PAK1 antibody produced in rabbit, affinity isolated antibody
SAB4502066
Anti-PAK1 antibody produced in rabbit, affinity isolated antibody
SAB4502063
Anti-PAK1, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB1406298
Anti-PKN1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA003982
Anti-PKN1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1411085
Anti-PKN1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1401681
Anti-PLEKHO1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
WH0001459M1 Monoclonal Anti-CSNK2A2 antibody produced in mouse, clone 4F2, purified immunoglobulin, buffered aqueous solution
SAB1403706
Monoclonal Anti-CSNK2A2, (N-terminal) antibody produced in mouse, clone 1E8, purified immunoglobulin, buffered aqueous solution
WH0005058M1
Monoclonal Anti-PAK1 antibody produced in mouse, clone 1E11, purified immunoglobulin, buffered aqueous solution
SAB1400193
Monoclonal Anti-PAK1 antibody produced in mouse, clone 4D1, purified immunoglobulin, buffered aqueous solution
WH0005585M1
Monoclonal Anti-PKN1 antibody produced in mouse, clone 1B10, purified immunoglobulin, buffered aqueous solution
SAB1404244
Monoclonal Anti-PKN1 antibody produced in mouse, clone 1A4, purified immunoglobulin, buffered aqueous solution
SAB1401682 Monoclonal Anti-PLEKHO1 antibody produced in mouse, clone 1C7, purified immunoglobulin, buffered aqueous solution
SAB1405085
Monoclonal Anti-PLEKHO1 antibody produced in mouse, clone 3D5, ascites fluid