EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Girdin/GIV is upregulated by cyclic tension, propagates mechanical signal transduction, and is required for the cellular proliferation and migration of MG-63 cells.
PMID 26163263
Abstract
To explore how Girdin/GIV is regulated by cyclic tension and propagates downstream signals to affect cell proliferation and migration. Human osteoblast-like MG-63 cells were exposed to cyclic tension force at 4000 μstrain and 0.5 Hz for 6 h, produced by a four-point bending system. Cyclic tension force upregulated Girdin and Akt expression and phosphorylation in cultured MG-63 cells. Girdin and Akt each promoted the phosphorylation of the other under stimulated tension. In vitro MTT and transwell assays showed that Girdin and Akt are required for cell proliferation and migration during cellular quiescence. Moreover, STAT3 was determined to be essential for Girdin expression under stimulated tension force in the physiological condition, as well as for osteoblast proliferation and migration during quiescence. These findings suggest that the STAT3/Girdin/Akt pathway activates in osteoblasts in response to mechanical stimulation and may play a significant role in triggering osteoblast proliferation and migration during orthodontic treatment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A4732
Anti-AKT2 (121-135) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
A4857
Anti-AKT2 (454-469) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB4300423
Anti-AKT2 (Ab-474) antibody produced in rabbit, affinity isolated antibody
SAB2501920
Anti-AKT2 (C-terminal) antibody produced in goat, affinity isolated antibody, buffered aqueous solution
GW22776 Anti-AKT2 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1411919
ANTI-AKT2 antibody produced in mouse, clone 1B4, purified immunoglobulin, buffered aqueous solution
SAB1411920
ANTI-AKT2 antibody produced in mouse, clone 1D9, purified immunoglobulin, buffered aqueous solution
SAB1411921
ANTI-AKT2 antibody produced in mouse, clone 1F3, purified immunoglobulin, buffered aqueous solution
SAB2103239
Anti-AKT2 antibody produced in rabbit, affinity isolated antibody
SAB2104347
Anti-AKT2, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB4500808
Anti-AKT2, C-Terminal antibody produced in rabbit, affinity isolated antibody
HPA038101
Anti-CCDC88A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB3500157
Anti-Girdin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4502344
Anti-GIRDIN antibody produced in rabbit, affinity isolated antibody
WH0000208M1
Monoclonal Anti-AKT2 antibody produced in mouse, clone 1B3, purified immunoglobulin, buffered aqueous solution
SAB5300348 Monoclonal Anti-AKT2 antibody produced in mouse, clone 1B6, ascites fluid
SAB3701428
Monoclonal Anti-AKT2 antibody produced in rat, clone 16G11.E8, purified immunoglobulin
PLA0245
Rabbit anti-AKT2 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.