EMAIL THIS PAGE TO A FRIEND
Environmental science and pollution research international
Assessment of oxytetracycline and tetracycline antibiotics in manure samples in different cities of Khuzestan Province, Iran.
PMID 26165993
Abstract
Tetracyclines (TCs), a class of antibiotics with a broad spectrum, are the most frequently used antibiotics in animal production. The major concern is that the widespread use of the antibiotics may lead to the emergence of new strains of bacteria that are resistant to these antibiotics. The objective of this study was to determine the residual levels of oxytetracycline and tetracycline in 80 animal manure samples that were collected from the livestock and poultry feedlots in Khuzestan Province. The residual levels of the antibiotics in the samples were extracted by using solid-phase extraction (SPE) method and subsequently were measured by liquid chromatography. Recoveries from the spiked poultry manure samples ranged from 65 to 113% for tetracycline and 86 to 132% for oxytetracycline. Relative standard deviations of the recoveries were less than 5.7% within the same day. Method detection limit (MDL) measured for oxytetracycline and tetracycline in the manure were 0.011 and 0.01 mg/kg, respectively. Analysis of the collected 50 chickens and 30 cow manure samples showed that the highest concentration of tetracycline was related to Behbahan City (5.36 mg/kg) and the lowest concentration was detected for Ramhormoz (0.05 mg/kg). The highest and lowest concentrations of oxytetracycline were respectively observed for Behbahan (13.77 mg/kg) and Ramhormoz (0.047 mg/kg). Based on the results, in chicken manure, there was significant statistical difference between the residual TC concentrations among five cities (p(value) < 0.05). However, no significant relationship was observed between oxytetracyclin (OTC) residual concentrations among five cities (p(value) > 0.05).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E5134
Ethylenediaminetetraacetic acid disodium salt dihydrate, for electrophoresis, for molecular biology, 99.0-101.0% (titration)
C10H14N2Na2O8 · 2H2O
E1644
Ethylenediaminetetraacetic acid disodium salt dihydrate, BioUltra, 98.5-101.5%
C10H14N2Na2O8 · 2H2O
E6635
Ethylenediaminetetraacetic acid disodium salt dihydrate, Sigma Grade, plant cell culture tested, 98.5-101.5%
C10H14N2Na2O8 · 2H2O
E5513
Ethylenediaminetetraacetic acid disodium salt dihydrate, for electrophoresis, 98.5-101.5%
C10H14N2Na2O8 · 2H2O
E4884
Ethylenediaminetetraacetic acid disodium salt dihydrate, ACS reagent, 99.0-101.0%
C10H14N2Na2O8 · 2H2O
ED2SS
Ethylenediaminetetraacetic acid disodium salt dihydrate, reagent grade, 98.5-101.5% (titration)
C10H14N2Na2O8 · 2H2O
27285
Ethylenediaminetetraacetic acid disodium salt dihydrate, meets analytical specification of Ph. Eur., BP, USP, FCC, 99.0-101.0%
C10H14N2Na2O8 · 2H2O
2854
Ethylenediaminetetraacetic acid disodium salt solution, 2%, solution
C10H14N2Na2O8
E7889
Ethylenediaminetetraacetic acid disodium salt solution, for molecular biology, 0.5 M in H2O, DNase, RNase, NICKase and protease, none detected
C10H14N2Na2O8
75688
Oxalic acid, puriss. p.a., anhydrous, ≥99.0% (RT)
C2H2O4
658537
Oxalic acid, purified grade, 99.999% trace metals basis
C2H2O4
241172
Oxalic acid, ReagentPlus®, ≥99%
C2H2O4
194131
Oxalic acid, 98%
C2H2O4