EMAIL THIS PAGE TO A FRIEND
Medicine
Impaired Phenotype and Function of T Follicular Helper Cells in HIV-1-Infected Children Receiving ART.
PMID 26166114
Abstract
T follicular helper (Tfh) cells are important components in development of specific humoral immune responses; whether the number and biology of Tfh cells is impaired in HIV-1-infected children is not yet studied.The frequency, phenotype, and function of Tfh cells and B cells were determined in blood of HIV-1-infected children receiving antiretroviral therapy (ART) and age-matched controls. Flow cytometry was used to characterize the frequency of Tfh cells and B cell subsets. Cytokine expression was measured after in vitro activation of Tfh cells.A reduced frequency of memory Tfh cells (P < 0.001) was identified in HIV-1-infected children and, on these cells, a reduced expression of programmed death-1 (PD-1) and inducible T cell costimulator (ICOS) (P < 0.001 and P < 0.01). Upon activation, the capacity of Tfh cells to express IL-4, an important cytokine for B cell function, was impaired in HIV-1-infected children.B cell subpopulations in HIV-1-infected children displayed significant differences from the control group: the frequency of resting memory (RM) B cells was reduced (P < 0.01) whereas the frequency of exhausted memory B cells increased (P < 0.001). Interestingly, the decline of RM cells correlated with the reduction of memory Tfh cells (P = 0.02).Our study shows that function and phenotype of Tfh cells, pivotal cells for establishment of adaptive B cell responses, are impaired during HIV-1 infection in children. A consistent reduction of memory Tfh cells is associated with declined frequencies of RM B cells, creating a novel link between dysfunctional features of these cell types, major players in establishment of humoral immunity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1408550
Anti-BLR1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4300659
Anti-CD10 antibody produced in rabbit, affinity isolated antibody
SAB4503617
Anti-CD19, C-Terminal antibody produced in rabbit, affinity isolated antibody
C8974 Anti-CD27 antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB1405582
Anti-CD27 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
C3366 Anti-CXCR5 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA042432
Anti-CXCR5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1401658
Anti-ICOS antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1306778
ANTI-ICOS(C-TERMINAL) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB3500345
Anti-IL-21 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1407827
Anti-IL21 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1411185
Anti-IL21 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
AV54329
Anti-IL4 antibody produced in rabbit, affinity isolated antibody
SAB2103750
Anti-IL4 antibody produced in rabbit, affinity isolated antibody
GW22461F Anti-Interleukin-2 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
I8398 Anti-Interleukin-2 antibody produced in goat, IgG fraction of antiserum, lyophilized powder
I8273 Anti-Interleukin-2 antibody produced in goat, IgG fraction of antiserum, lyophilized powder
I7401 Anti-Interleukin-2 antibody produced in goat, affinity isolated antibody, lyophilized powder
I7784 Anti-Interleukin-2 antibody produced in goat, affinity isolated antibody, lyophilized powder
GW21034 Anti-Interleukin-21 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
GW22812 Anti-Interleukin-21 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
I8533 Anti-Interleukin-21 antibody produced in goat, affinity isolated antibody, lyophilized powder
I7776 Anti-Interleukin-4 antibody produced in goat, affinity isolated antibody, lyophilized powder
I7526 Anti-Interleukin-4 antibody produced in goat, affinity isolated antibody, lyophilized powder
I8784 Anti-Interleukin-4 antibody produced in goat, IgG fraction of antiserum, lyophilized powder
SAB1400166
Anti-MME antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
SAB1401246
Anti-MME antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
PRS4065
Anti-PD-1 (ab1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS4067
Anti-PD-1 (ab2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2900280
Anti-PDCD1 (AB1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution