EMAIL THIS PAGE TO A FRIEND
Journal of pediatric neurosciences
A rare case of glycine encephalopathy unveiled by valproate therapy.
PMID 26167219
Abstract
Glycine encephalopathy (GE) or nonketotic hyperglycinemia is an autosomal recessive disorder due to a primary defect in glycine cleavage enzyme system. It is characterized by elevated levels of glycine in plasma and cerebrospinal fluid usually presenting with seizures, hypotonia, and developmental delay. In our case, paradoxical increase in seizure frequency on starting sodium valproate led us to diagnose GE.