Description
Molecular Formula
Add to Cart
SAB4300464
Anti-APP (Ab-668) antibody produced in rabbit, affinity isolated antibody
SAB2100134
Anti-APP (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2100135
Anti-APP (ab2) antibody produced in rabbit, affinity isolated antibody
SAB2100136
Anti-APP (ab3) antibody produced in rabbit, IgG fraction of antiserum
SAB2502012
Anti-APP (N-terminal) antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA001462
Anti-APP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4200535
Anti-APP antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
SAB3500274
Anti-APP antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB5200113
Anti-APP antibody produced in rabbit, 1 mg/mL, affinity isolated antibody
SAB2107345
Anti-APP antibody produced in rabbit, affinity isolated antibody
SAB2106270
Anti-APP antibody produced in rabbit, affinity isolated antibody
WH0000351M1
Monoclonal Anti-APP antibody produced in mouse, clone 2C12, purified immunoglobulin, buffered aqueous solution
SAB4100478
Monoclonal Anti-APP antibody produced in mouse, clone APP-A434, culture supernatant
SAB4100500
Monoclonal Anti-APP antibody produced in mouse, clone APP-B434, culture supernatant
SAB1403560
Monoclonal Anti-APP, (C-terminal) antibody produced in mouse, clone 1E6, purified immunoglobulin, buffered aqueous solution