EMAIL THIS PAGE TO A FRIEND
The Journal of investigative dermatology
Defining the Contribution of MC1R Physiological Ligands to ATR Phosphorylation at Ser435, a Predictor of DNA Repair in Melanocytes.
PMID 26168232
Abstract
The melanocortin 1 receptor (MC1R), a GS-coupled receptor that signals through cAMP and protein kinase A (PKA), regulates pigmentation, adaptive tanning, and melanoma resistance. MC1R-cAMP signaling promotes PKA-mediated phosphorylation of ataxia telangiectasia and rad3-related (ATR) at Ser435 (ATR-pS435), a modification that enhances nucleotide excision repair (NER) by facilitating recruitment of the XPA protein to sites of UV-induced DNA damage. High-throughput methods were developed to quantify ATR-pS435, measure XPA-photodamage interactions, and assess NER function. We report that melanocyte-stimulating hormone (α-MSH) or ACTH induce ATR-pS435, enhance XPA's association with UV-damaged DNA and optimize melanocytic NER. In contrast, MC1R antagonists agouti signaling protein (ASIP) or human β-defensin 3 (HBD3) interfere with ATR-pS435 generation, impair the XPA-DNA interaction, and reduce DNA repair. Although ASIP and HBD3 each blocked α-MSH-mediated induction of the signaling pathway, only ASIP depleted basal ATR-pS435. Our findings confirm that ASIP diminishes agonist-independent MC1R basal signaling whereas HBD3 is a neutral MC1R antagonist that blocks activation by melanocortins. Furthermore, our data suggest that ATR-pS435 may be a useful biomarker for the DNA repair-deficient MC1R phenotype.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2100125
Anti-ANTXR1 antibody produced in rabbit, affinity isolated antibody
HPA052046
Anti-ANTXR1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4300576
Anti-ATR (Ab-428) antibody produced in rabbit, affinity isolated antibody
PRS3117
Anti-ATR (ab1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS3119
Anti-ATR (ab2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4200348
Anti-ATR (C-terminal) antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
HPA028264
Anti-ATR antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1305004
ANTI-CPD (C-TERM) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
AV53585
Anti-CPD antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB2700486
Anti-CPD antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA052796
Anti-CPD antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1400346
Anti-HOMER2 antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
HPA040134
Anti-HOMER2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1411660
Anti-HOMER2 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1409213
Anti-MMAB antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA039017
Anti-MMAB antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4503908
Anti-phospho-CREB (pSer121) antibody produced in rabbit, affinity isolated antibody
SAB4504375
Anti-phospho-CREB (pSer133) antibody produced in rabbit, affinity isolated antibody
SAB4503757
Anti-phospho-CREB (pSer142) antibody produced in rabbit, affinity isolated antibody
SAB4504577
Anti-phospho-CREB (pThr100) antibody produced in rabbit, affinity isolated antibody
SAB1306301 ANTI-PHOSPHO-CREB(S133) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4300217
Anti-phospho-CREB1 (pSer129) antibody produced in rabbit, affinity isolated antibody
SAB4300040
Anti-phospho-CREB1 (pSer133) antibody produced in rabbit, affinity isolated antibody
X1254 Anti-XPA (C-terminal) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
X1504 Anti-XPA (N-terminal) antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB1406599
Anti-XPA antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1411400
Anti-XPA antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4503090
Anti-XPA, C-Terminal antibody produced in rabbit, affinity isolated antibody
WH0000545M1
Monoclonal Anti-ATR antibody produced in mouse, clone 3F2, purified immunoglobulin, buffered aqueous solution
SAB1403593
Monoclonal Anti-ATR, (C-terminal) antibody produced in mouse, clone 1E9, purified immunoglobulin, buffered aqueous solution