Description
Molecular Formula
Add to Cart
PHR1800 Metamizole Sodium, Pharmaceutical Secondary Standard; Certified Reference Material
C13H16N3O4S · Na · H2O
SML1488 Metamizole sodium hydrate, ≥98% (HPLC)
C13H16N3O4S · Na · H2O