EMAIL THIS PAGE TO A FRIEND
Journal of immunology (Baltimore, Md. : 1950)
Oil-in-Water Emulsion MF59 Increases Germinal Center B Cell Differentiation and Persistence in Response to Vaccination.
PMID 26170383
Abstract
Induction of persistent protective immune responses is a key attribute of a successful vaccine formulation. MF59 adjuvant, an oil-in-water emulsion used in human vaccines, is known to induce persistent high-affinity functional Ab titers and memory B cells, but how it really shapes the Ag-specific B cell compartment is poorly documented. In this study, we characterized the Ab- and Ag-specific B cell compartment in wild-type mice immunized with HlaH35L, a Staphylococcus aureus Ag known to induce measurable functional Ab responses, formulated with MF59 or aluminum salts, focusing on germinal centers (GC) in secondary lymphoid organs. Taking advantage of single-cell flow cytometry analyses, HlaH35L-specific B cells were characterized for the expression of CD38 and GL-7, markers of memory and GC, respectively, and for CD80 and CD73 activation markers. We demonstrated that immunization with MF59-, but not aluminum salt-adjuvanted HlaH35L, induced expanded Ag-specific CD73(+)CD80(-) GC B cells in proximal- and distal-draining lymph nodes, and promoted the persistence of GC B cells, detected up to 4 mo after immunization. In addition to increasing GC B cells, MF59-adjuvanted HlaH35L also increased the frequency of T follicular helper cells. This work extends previous knowledge regarding adaptive immune responses to MF59-adjuvanted vaccines, and, to our knowledge, for the first time an adjuvant used in human licensed products is shown to promote strong and persistent Ag-specific GC responses that might benefit the rational design of new vaccination strategies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GW22573F Anti-BB1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1408550
Anti-BLR1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2501656
Anti-CD80 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1405584
Anti-CD80 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
C3366 Anti-CXCR5 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA042432
Anti-CXCR5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1401658
Anti-ICOS antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1306778
ANTI-ICOS(C-TERMINAL) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1406012
Anti-IGHD antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA017357
Anti-NT5E antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
WH0000643M1
Monoclonal Anti-BLR1 antibody produced in mouse, clone 2C1, purified immunoglobulin, buffered aqueous solution
SAB4700245
Monoclonal Anti-CD21 antibody produced in mouse, clone LT21, purified immunoglobulin, buffered aqueous solution
WH0000941M1 Monoclonal Anti-CD80 antibody produced in mouse, clone 1G1, purified immunoglobulin, buffered aqueous solution
SAB1403647
Monoclonal Anti-CD80 antibody produced in mouse, clone 3E10, ascites fluid
SAB4700140 Monoclonal Anti-CD80 antibody produced in mouse, clone MEM-233, purified immunoglobulin, buffered aqueous solution
WH0029851M1 Monoclonal Anti-ICOS antibody produced in mouse, clone 1G1, purified immunoglobulin, buffered aqueous solution
SAB1412835
Monoclonal Anti-ICOS antibody produced in mouse, clone 3G4, purified immunoglobulin, 0.5 mg/mL
I2768 Monoclonal Anti-IgD antibody produced in mouse, clone HJ9, purified immunoglobulin, buffered aqueous solution
WH0004907M1
Monoclonal Anti-NT5E antibody produced in mouse, clone 4C4-2B5, purified immunoglobulin, buffered aqueous solution
SAB5300408 Monoclonal Anti-NT5E antibody produced in mouse, clone 1D7, ascites fluid
SAB1404149
Monoclonal Anti-NT5E, (C-terminal) antibody produced in mouse, clone 4D3-2C10, purified immunoglobulin, buffered aqueous solution
SAB1402533
Monoclonal Anti-NTE, (C-terminal) antibody produced in mouse, clone 3D10, purified immunoglobulin, buffered aqueous solution
S3626
Squalene, ≥98%, liquid
C30H50