EMAIL THIS PAGE TO A FRIEND
Oncology letters
Overexpression and delocalization of claudin-3 protein in MCF-7 and MDA-MB-415 breast cancer cell lines.
PMID 26170992
Abstract
Tumor-specific deregulated expression of claudins, integral membrane proteins found in tight junctions (TJs), has indicated a possible role for TJ disruption in cancer progression. The current study demonstrates the marked overexpression of claudin-3 protein in two breast cancer cell lines of metastatic origin (MCF-7 and MDA-MB-415). Immunofluorescence and differential detergent fractionation analyses revealed that, although claudin-3 was primarily localized at cell junctions, it was also detected intracellularly. Similarly, the siRNA-mediated suppression of claudin-3 did not considerably affect its pattern of subcellular distribution relative to mock-transfected cells. However, there appeared to be a preferential loss of claudin-3 signal in the cytoskeletal fraction. Wound-healing assays were conducted to assess the effect of endogenous overexpression versus siRNA-mediated suppression of claudin-3 on cellular motility in MCF-7 cells. Suppression of claudin-3 protein levels resulted in a marked decrease in the rate of cellular motility relative to mock-transfected cells. These findings suggest that overexpression of claudin-3 may be important in disrupting TJ integrity and thus contribute to enhanced cellular motility, a key component of tumor progression.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4503545
Anti-Claudin 1, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4503546
Anti-Claudin 1, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4500434
Anti-Claudin 3, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4500435
Anti-Claudin 3, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4500436
Anti-Claudin 7, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4500437
Anti-Claudin 7, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4200534
Anti-Claudin-1 (C-terminal) antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
SAB4200573
Anti-Claudin-4 antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
HPA014361 Anti-CLDN3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2106604
Anti-CLDN3 antibody produced in rabbit, affinity isolated antibody
SAB1408889
Monoclonal Anti-CLDN3 antibody produced in mouse, clone 2F2, purified immunoglobulin, buffered aqueous solution
WH0001364M1
Monoclonal Anti-CLDN4 antibody produced in mouse, clone 4A4, purified immunoglobulin, buffered aqueous solution
SAB1401080
Monoclonal Anti-CLDN4 antibody produced in mouse, clone 4A4, ascites fluid
SAB1403689
Monoclonal Anti-CLDN4 antibody produced in mouse, clone 4A11, purified immunoglobulin, buffered aqueous solution