EMAIL THIS PAGE TO A FRIEND
Beilstein journal of nanotechnology
The convenient preparation of stable aryl-coated zerovalent iron nanoparticles.
PMID 26171295
Abstract
A novel approach for the in situ synthesis of zerovalent aryl-coated iron nanoparticles (NPs) based on diazonium salt chemistry is proposed. Surface-modified zerovalent iron NPs (ZVI NPs) were prepared by simple chemical reduction of iron(III) chloride aqueous solution followed by in situ modification using water soluble arenediazonium tosylate. The resulting NPs, with average iron core diameter of 21 nm, were coated with a 10 nm thick organic layer to provide long-term protection in air for the highly reactive zerovalent iron core up to 180 °C. The surface-modified iron NPs possess a high grafting density of the aryl group on the NPs surface of 1.23 mmol/g. FTIR spectroscopy, XRD, HRTEM, TGA/DTA, and elemental analysis were performed in order to characterize the resulting material.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
235385
tert-Butyl nitrite, 90%
C4H9NO2