EMAIL THIS PAGE TO A FRIEND
British journal of cancer
Understanding cervical screening non-attendance among ethnic minority women in England.
PMID 26171938
Abstract
Women from Black, Asian and Minority Ethnic (BAME) backgrounds are less likely to attend cervical screening than White British women. This study explored sociodemographic and attitudinal correlates of cervical screening non-attendance among BAME women. Women (30-60 years) were recruited from Indian, Pakistani, Bangladeshi, Caribbean, African and White British backgrounds (n=720). Participants completed structured interviews. BAME women were more likely to be non-attenders than white British women (44-71% vs 12%) and fell into two groups: the disengaged and the overdue. Migrating to the United Kingdom, speaking a language other than English and low education level were associated with being disengaged. Being overdue was associated with older age. Three attitudinal barriers were associated with being overdue for screening among BAME women: low perceived risk of cervical cancer due to sexual inactivity, belief that screening is unnecessary without symptoms and difficulty finding an appointment that fits in with other commitments. BAME non-attenders appear to fall into two groups, and interventions for these groups may need to be targeted and tailored accordingly. It is important to ensure that BAME women understand cancer screening is intended for asymptomatic women and those who have ceased sexual activity may still be at risk.