EMAIL THIS PAGE TO A FRIEND
Polish journal of veterinary sciences
Yersinia enterocolitica strains isolated from beavers (Castor fiber).
PMID 26172198
Abstract
Pseudocloacal swabs and palatine tonsils from beavers have been examined for the Yersinia enterocolitica presence. Thirty-six samples from 24 beavers were collected and subjected to bacteriological examinations including sero- and biotypisation. Amplicons confirmed by PCR as Y. enterocolitica were sequenced. Positive samples originated from 4 out of the 24 beavers (16.7 %) and all the strains belonged to biotype 1A. The study suggested that Y. enterocolitica could be isolated from beavers, which may therefore be treated as a reservoir, a significant factor of water contamination and a vector of the Y. enterocolitica.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P1338
Potassium chlorate, BioXtra, ≥99.0%
ClKO3