EMAIL THIS PAGE TO A FRIEND
Meat science
Evaluation of combined effects of ageing period and freezing rate on quality attributes of beef loins.
PMID 26172242
Abstract
The objective of our study was to evaluate the combined effects of ageing period and different freezing rates on meat quality attributes of beef loins. Pairs of loins (M. longissimus at 1 day post mortem) from 12 carcasses were divided into four equal portions and randomly assigned to four ageing/freezing treatments (aged only, frozen only, and 3 or 4 weeks ageing at -1.5°C then frozen). Two freezing methods (fast freezing by calcium chloride immersion or slow freezing by air freezer at -18°C) were applied to the loin sections. Fast freezing had no effect on shear force (P>0.05), but significantly improved the water-holding capacity of the aged/frozen loins by reducing purge and drip losses. Ageing-then-freezing significantly improved shear force values of loins compared to both the aged only and frozen only loins. These observations suggest that fast freezing will add more value to the aged/frozen/thawed meat by minimising the amount of water-loss due to the freezing/thawing process.