EMAIL THIS PAGE TO A FRIEND
Neurobiology of aging
The amyloid precursor protein (APP) intracellular domain regulates translation of p44, a short isoform of p53, through an IRES-dependent mechanism.
PMID 26174856
Abstract
p44 is a short isoform of the tumor suppressor protein p53 that is regulated in an age-dependent manner. When overexpressed in the mouse, it causes a progeroid phenotype that includes premature cognitive decline, synaptic defects, and hyperphosphorylation of tau. The hyperphosphorylation of tau has recently been linked to the ability of p44 to regulate transcription of relevant tau kinases. Here, we report that the amyloid precursor protein (APP) intracellular domain (AICD), which results from the processing of the APP, regulates translation of p44 through a cap-independent mechanism that requires direct binding to the second internal ribosome entry site (IRES) of the p53 mRNA. We also report that AICD associates with nucleolin, an already known IRES-specific trans-acting factor that binds with p53 IRES elements and regulates translation of p53 isoforms. The potential biological impact of our findings was assessed in a mouse model of Alzheimer's disease. In conclusion, our study reveals a novel aspect of AICD and p53/p44 biology and provides a possible molecular link between APP, p44, and tau.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4300464
Anti-APP (Ab-668) antibody produced in rabbit, affinity isolated antibody
SAB2100134
Anti-APP (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2100135
Anti-APP (ab2) antibody produced in rabbit, affinity isolated antibody
SAB2100136
Anti-APP (ab3) antibody produced in rabbit, IgG fraction of antiserum
SAB2502012
Anti-APP (N-terminal) antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA001462
Anti-APP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4200535
Anti-APP antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
SAB3500274
Anti-APP antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB5200113
Anti-APP antibody produced in rabbit, 1 mg/mL, affinity isolated antibody
SAB2107345
Anti-APP antibody produced in rabbit, affinity isolated antibody
SAB2106270
Anti-APP antibody produced in rabbit, affinity isolated antibody
SAB4300374
Anti-MAPT (Ab-181) antibody produced in rabbit, affinity isolated antibody
SAB4300375
Anti-MAPT (Ab-205) antibody produced in rabbit, affinity isolated antibody
SAB4300503
Anti-MAPT (Ab-212) antibody produced in rabbit, affinity isolated antibody
SAB4300376
Anti-MAPT (Ab-214) antibody produced in rabbit, affinity isolated antibody
SAB4300377
Anti-MAPT (Ab-231) antibody produced in rabbit, affinity isolated antibody
SAB4300373
Anti-MAPT (Ab-235) antibody produced in rabbit, affinity isolated antibody
SAB4300378
Anti-MAPT (Ab-262) antibody produced in rabbit, affinity isolated antibody
SAB4300370
Anti-MAPT (Ab-356) antibody produced in rabbit, affinity isolated antibody
SAB4300371
Anti-MAPT (Ab-396) antibody produced in rabbit, affinity isolated antibody
SAB4300379
Anti-MAPT (Ab-404) antibody produced in rabbit, affinity isolated antibody
SAB4300516
Anti-MAPT (Ab-422) antibody produced in rabbit, affinity isolated antibody
AV48103
Anti-MAPT antibody produced in rabbit, affinity isolated antibody
SAB4501821
Anti-TAU antibody produced in rabbit, affinity isolated antibody
SAB4501827
Anti-TAU antibody produced in rabbit, affinity isolated antibody
SAB4501828
Anti-TAU antibody produced in rabbit, affinity isolated antibody
SAB4501829
Anti-TAU antibody produced in rabbit, affinity isolated antibody
SAB4501831
Anti-TAU antibody produced in rabbit, affinity isolated antibody
SAB4501832
Anti-TAU antibody produced in rabbit, affinity isolated antibody
SAB1306711
ANTI-TAU(C-TERMINAL) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution