EMAIL THIS PAGE TO A FRIEND
Journal of biomedical science
Role of Raf-kinase inhibitor protein in colorectal cancer and its regulation by hydroxycamptothecine.
PMID 26177829
Abstract
Recently accumulated evidence suggests that Raf kinase inhibitor protein (RKIP) participates in regulation of many signaling pathways and plays an important role in tumorigenesis and tumor metastasis. However, studies investigating the role of RKIP in colorectal cancer have not been reported. The aim of this study was to investigate the role of RKIP on colorectal cancer cell differentiation, progression and its correlation with chemosensitivity. Immunohistochemical analysis revealed that RKIP expression was higher in non-neoplastic colorectal tissue (NCRCT) and colorectal cancer tissue (CRCT) than that in metastatic lymph node tissue (MLNT) (P <0.05). P-ERK protein expression was higher in MLNT and CRCT than that in NCRCT (P = 0.02). Immunocytochemical analysis further revealed that RKIP expression was higher in the well differentiated cell line SW1116 as compared to that in the poorly differentiated cell line LoVo. Matrigel invasive assay demonstrated that the inhibition of RKIP by short hairpin RNA (shRNA) 271 transfection significantly increased the number of migrated cells (90.67 ± 4.04 vs. 37.33 ± 2.51, P <0.05), whereas over-expression of RKIP by PEBP-1 plasmid transfection significantly suppressed the number of migrated cells (79.24 ± 5.18 vs. 154.33 ± 7.25, P <0.05). Meanwhile, down-regulation of RKIP induced an increase in the cell survival rate by inhibiting apoptosis induced by hydroxycamptothecine. RKIP was also found to be associated with cell differentiation, with a higher activity in well differentiated colorectal cancer cells than in poorly differentiated ones. The upregulated expression of RKIP in colorectal cancer cells inhibited cell invasion and metastasis, while downregulation of RKIP reduced chemosensitivity by inhibiting apoptosis induced by HCPT.