EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim an der Bergstrasse, Germany)
Selective Single-Step Separation of a Mixture of Three Metal Ions by a Triphasic Ionic-Liquid-Water-Ionic-Liquid Solvent Extraction System.
PMID 26178665
Abstract
In a conventional solvent extraction system, metal ions are distributed between two immiscible phases, typically an aqueous and an organic phase. In this paper, the proof-of-principle is given for the distribution of metal ions between three immiscible phases, two ionic liquid phases with an aqueous phase in between them. Three-liquid-phase solvent extraction allows separation of a mixture of three metal ions in a single step, whereas at least two steps are required to separate three metals in the case of two-liquid-phase solvent extraction. In the triphasic system, the lower organic phase is comprised of the ionic liquid betainium- or choline bis(trifluoromethylsulfonyl)imide, whereas the upper organic phase is comprised of the ionic liquid trihexyl(tetradecyl)phosphonium bis(trifluoromethylsulfonyl)imide. The triphasic system was used for the separation of a mixture of tin(II), yttrium(III), and scandium(III) ions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B2629
Betaine, ≥98% (perchloric acid titration)
C5H11NO2
61962
Betaine, BioUltra, ≥99.0% (NT)
C5H11NO2
B0300
Betaine solution, 5 M, PCR Reagent
C5H11NO2
18472
Betaine solution, 0.1 M
C5H11NO2
461210
Cerium, chips
Ce
261041
Cerium, ingot, under oil, 99.9% trace rare earth metals basis
Ce
263001
Cerium, powder, −40 mesh, 99.9% trace rare earth metals basis
Ce
GF97217220
Cerium, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99.9%
Ce
GF49222582
Cerium, wire reel, 25mm, diameter 1.0mm, hard, 99.9%
Ce
GF61818363
Cerium, foil, 25x25mm, thickness 0.1mm, as rolled, 99.9%
Ce
GF27295724
Cerium, foil, 50x50mm, thickness 0.1mm, as rolled, 99.9%
Ce
GF97378677
Cerium, foil, light tested, 25x25mm, thickness 0.25mm, as rolled, 99.9%
Ce
GF36157394
Cerium, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99.9%
Ce
GF90687695
Cerium, wire reel, 50mm, diameter 1.0mm, hard, 99.9%
Ce
GF39030353
Cerium, rod, 100mm, diameter 12.7mm, cast, 99.9%
Ce
GF04444095
Cerium, rod, 100mm, diameter 6.35mm, cast, 99.9%
Ce
GF55512244
Cerium, rod, 50mm, diameter 12.7mm, cast, 99.9%
Ce
GF02435554
Cerium, rod, 50mm, diameter 6.35mm, cast, 99.9%
Ce
GF35961655
Cerium, wire reel, 100mm, diameter 1.0mm, hard, 99.9%
Ce
GF49695047
Cerium, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99.9%
Ce
C7527
Choline chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
C5H14ClNO
C7017
Choline chloride, ≥99%
C5H14ClNO
C1879
Choline chloride, ≥98%
C5H14ClNO
26978
Choline chloride, BioUltra, ≥99.0% (AT)
C5H14ClNO
295345
Helium, ≥99.995%
He
225827
N,N′-Disuccinimidyl carbonate, ≥95%
C9H8N2O7
158615
Tributyl phosphate, 97%
C12H27O4P
240494
Tributyl phosphate, ≥99%
C12H27O4P
T7567
Tributylphosphine solution, 200 mM (in N-methyl-2-pyrrolidinone), liquid
C12H27P