EMAIL THIS PAGE TO A FRIEND
The American journal of clinical nutrition
The 2-monoacylglycerol moiety of dietary fat appears to be responsible for the fat-induced release of GLP-1 in humans.
PMID 26178726
Abstract
Dietary triglycerides can, after digestion, stimulate the intestinal release of incretin hormones through activation of G protein-coupled receptor (GPR) 119 by 2-monoacylglycerol and by the activation of fatty acid receptors for long- and short-chain fatty acids. Medium-chain fatty acids do not stimulate the release of intestinal hormones. To dissect the mechanism of fat-induced glucagon-like peptide 1 (GLP-1) and glucose-dependent insulinotropic polypeptide (GIP) release in humans, we compared the effects of tributyrin (containing short-chain fatty acids; i.e., butyric acid), olive oil [containing long-chain fatty acids; e.g., oleic acid plus 2-oleoyl glycerol (2-OG)], and 1,3-dioctanoyl-2-oleoyl glycerol (C8-dietary oil), which is digested to form medium-chain fatty acids : i.e., octanoic acid : and 2-OG. In a randomized, single-blinded crossover study, 12 healthy white men [mean age: 24 y; BMI (in kg/m(2)): 22] were given the following 4 meals on 4 different days: 200 g carrots + 6.53 g tributyrin, 200 g carrots + 13.15 g C8-dietary oil, 200 g carrots + 19 g olive oil, or 200 g carrots. All of the lipids totaled 0.0216 mol. Main outcome measures were incremental areas under the curve for total GLP-1, GIP, and cholecystokinin (CCK) in plasma. C8-dietary oil and olive oil showed the same GLP-1 response [583 ± 101 and 538 ± 71 (pmol/L) × 120 min; P = 0.733], whereas the GIP response was higher for olive oil than for C8-dietary oil [3293 ± 404 and 1674 ± 270 (pmol/L) × 120 min; P = 0.002]. Tributyrin and carrots alone resulted in no increase in any of the measured hormones. Peptide YY (PYY) and neurotensin responses resembled those of GLP-1. Only olive oil stimulated CCK release. Under our study conditions, 2-OG and GPR119 activation can fully explain the olive oil-induced secretion of GLP-1, PYY, and neurotensin. In contrast, both oleic acid and 2-OG contributed to the GIP response. Dietary butyrate did not stimulate gut hormone secretion. Olive oil-derived oleic acid seems to be fully responsible for olive oil-induced CCK secretion. This trial was registered at clinicaltrials.gov as NCT02264951.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
TR0100 Serum Triglyceride Determination Kit,  kit sufficient for 250 tests