EMAIL THIS PAGE TO A FRIEND
The American journal of clinical nutrition
High provitamin A carotenoid serum concentrations, elevated retinyl esters, and saturated retinol-binding protein in Zambian preschool children are consistent with the presence of high liver vitamin A stores.
PMID 26178727
Abstract
Biomarkers of micronutrient status are needed to best define deficiencies and excesses of essential nutrients. We evaluated several supporting biomarkers of vitamin A status in Zambian children to determine whether any of the biomarkers were consistent with high liver retinol stores determined by using retinol isotope dilution (RID). A randomized, placebo-controlled, biofortified maize efficacy trial was conducted in 140 rural Zambian children from 4 villages. A series of biomarkers were investigated to better define the vitamin A status of these children. In addition to the assessment of total-body retinol stores (TBSs) by using RID, tests included analyses of serum carotenoids, retinyl esters, and pyridoxal-5'-phosphate (PLP) by using high-pressure liquid chromatography, retinol-binding protein by using ELISA, and alanine aminotransferase (ALT) activity by using a colorimetric assay. Children (n = 133) were analyzed quantitatively for TBSs by using RID. TBSs, retinyl esters, some carotenoids, and PLP differed by village site. Serum carotenoids were elevated above most nonintervened reference values for children. α-Carotene, β-carotene, and lutein values were >95th percentile from children in the US NHANES III, and 13% of children had hypercarotenemia (defined as total carotenoid concentration >3.7 μmol/L). Although only 2% of children had serum retinyl esters >10% of total retinol plus retinyl esters, 16% of children had >5% as esters, which was consistent with high liver retinol stores. Ratios of serum retinol to retinol-binding protein did not deviate from 1.0, which indicated full saturation. ALT activity was low, which was likely due to underlying vitamin B-6 deficiency, which was confirmed by very low serum PLP concentrations. The finding of hypervitaminosis A in Zambian children was supported by high circulating concentrations of carotenoids and mildly elevated serum retinyl esters. ALT-activity assays may be compromised with co-existing vitamin B-6 deficiency. Nutrition education to improve intakes of whole grains and animal-source foods may enhance vitamin B-6 status in Zambians.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A7502
DL-Alanine, ≥99% (HPLC)
C3H7NO2
W381810
DL-Alanine, ≥99%, FCC, FG
C3H7NO2