EMAIL THIS PAGE TO A FRIEND
The Journal of neuroscience : the official journal of the Society for Neuroscience
mRNAs and Protein Synthetic Machinery Localize into Regenerating Spinal Cord Axons When They Are Provided a Substrate That Supports Growth.
PMID 26180210
Abstract
Although intra-axonal protein synthesis is well recognized in cultured neurons and during development in vivo, there have been few reports of mRNA localization and/or intra-axonal translation in mature CNS axons. Indeed, previous work indicated that mature CNS axons contain much lower quantities of translational machinery than PNS axons, leading to the conclusion that the capacity for intra-axonal protein synthesis is linked to the intrinsic capacity of a neuron for regeneration, with mature CNS neurons showing much less growth after injury than PNS neurons. However, when regeneration by CNS axons is facilitated, it is not known whether the intra-axonal content of translational machinery changes or whether mRNAs localize into these axons. Here, we have used a peripheral nerve segment grafted into the transected spinal cord of adult rats as a supportive environment for regeneration by ascending spinal axons. By quantitative fluorescent in situ hybridization combined with immunofluorescence to unambiguously distinguish intra-axonal mRNAs, we show that regenerating spinal cord axons contain β-actin, GAP-43, Neuritin, Reg3a, Hamp, and Importin β1 mRNAs. These axons also contain 5S rRNA, phosphorylated S6 ribosomal protein, eIF2α translation factor, and 4EBP1 translation factor inhibitory protein. Different levels of these mRNAs in CNS axons from regenerating PNS axons may relate to differences in the growth capacity of these neurons, although the presence of mRNA transport and likely local translation in both CNS and PNS neurons suggests an active role in the regenerative process. Although peripheral nerve axons retain the capacity to locally synthesize proteins into adulthood, previous studies have argued that mature brain and spinal cord axons cannot synthesize proteins. Protein synthesis in peripheral nerve axons is increased during regeneration, and intra-axonally synthesized proteins have been shown to contribute to nerve regeneration. Here, we show that mRNAs and translational machinery are transported into axons regenerating from the spinal cord into the permissive environment of a peripheral nerve graft. Our data raise the possibility that spinal cord axons may make use of localized protein synthesis for regeneration.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB3500210
Anti-4E-BP1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4500736
Anti-4E-BP1 antibody produced in rabbit, affinity isolated antibody
SAB4500737
Anti-4E-BP1 antibody produced in rabbit, affinity isolated antibody
SAB4300474
Anti-EIF4EBP1 (Ab-36) antibody produced in rabbit, affinity isolated antibody
SAB4300475
Anti-EIF4EBP1 (Ab-45) antibody produced in rabbit, affinity isolated antibody
HPA023501
Anti-EIF4EBP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2106955
Anti-EIF4EBP1 antibody produced in rabbit, affinity isolated antibody
SAB1406176
Anti-NEFL antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1408610
Anti-NEFL antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2501318
Anti-NEFM antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA022845
Anti-NEFM antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
HPA023138
Anti-NEFM antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
SAB2501861
Anti-NFASC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA008832 Anti-NFASC antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA002044
Anti-PSMC4 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA005471
Anti-PSMC4 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2105008
Anti-PSMC4, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB2105009
Anti-PSMC4, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB4300484
Anti-RPS6 (Ab-235) antibody produced in rabbit, affinity isolated antibody
SAB1300250
Anti-RPS6 (N-term) antibody produced in rabbit, Ig fraction of antiserum, buffered aqueous solution
HPA031153
Anti-RPS6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2107629
Anti-RPS6 antibody produced in rabbit, affinity isolated antibody
WH0001978M1
Monoclonal Anti-EIF4EBP1 antibody produced in mouse, clone 4F3-H2, purified immunoglobulin, buffered aqueous solution
SAB1403761
Monoclonal Anti-EIF4EBP1 antibody produced in mouse, clone 1F7, purified immunoglobulin, buffered aqueous solution
SAB5300484
Monoclonal Anti-EIF4EBP1 antibody produced in mouse, clone 9E12D9, ascites fluid
SAB5300486
Monoclonal Anti-EIF4EBP1 antibody produced in mouse, clone 11G12C11, ascites fluid
WH0004744M1
Monoclonal Anti-NEFH antibody produced in mouse, clone 2E8, purified immunoglobulin, buffered aqueous solution
SAB5300413
Monoclonal Anti-NEFL antibody produced in mouse, clone 1H3, ascites fluid
SAB5300414
Monoclonal Anti-NEFL antibody produced in mouse, clone 2G10, purified immunoglobulin, buffered aqueous solution
SAB4100574
Monoclonal Anti-NEFL antibody produced in mouse, clone NEFL-A546, culture supernatant