EMAIL THIS PAGE TO A FRIEND
Zeitschrift fur Naturforschung. C, Journal of biosciences
Feeding stimulants for larvae of Graphium sarpedon nipponum (Lepidoptera: Papilionidae) from Cinnamomum camphora.
PMID 26181048
Abstract
The feeding response of larvae of the swallowtail butterfly, Graphium sarpedon nipponum (Lepidoptera: Papilionidae), is elicited by a methanolic extract from camphor tree (Cinnamomum camphora) leaves. Based on bioassay-guided fractionation, three compounds, isolated from the methanolic extract of fresh leaves of the camphor tree, were revealed to be involved in a multi-component system of feeding stimulants. Structures of these feeding stimulants were identified as sucrose, 5-O-caffeoylquinic acid and quercetin 3-O-β-glucopyranoside by NMR and LC-MS.